Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive test controls include: PC-12, U-251MG. The target is usually found in the following locations: Nucleus.
Immunogen sequence: MYKMEYSYLN SSAYESCMAG MDTSSLASAY ADFSSCSQAS GFQYNPIRTT FGATSGCPSL TPGSCSLGTL RDHQSSPYAA VPYKLFTDHG
The DNA-associated protein encoded by this gene is a member of the paired family of homeobox proteins localized to the nucleus. The protein functions as a transcription factor involved in the development of several major noradrenergic neuron populations and the determination of neurotransmitter phenotype. The gene product is linked to enhancement of second messenger-mediated activation of the dopamine beta-hydroylase, c-fos promoters and several enhancers, including cyclic amp-response element and serum-response element.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dilated pupils 1; GENA 269; NBPhox; neuroblastoma paired-type homeobox protein; Neuroblastoma Phox; paired mesoderm homeobox 2b; Paired mesoderm homeobox protein 2B; Paired-like homeobox 2B; PHOX2B homeodomain protein
Gene Aliases: Dilp1; NBLST2; NBPhox; PHOX2B; PMX2B
UniProt ID: (Human) Q99453, (Mouse) O35690
Entrez Gene ID: (Human) 8929, (Rat) 364152, (Mouse) 18935
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support