Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MEQLRAAARL QIVLGHLGRP SAGAVVAHPT SGTISSASFH PQQFQYTLDN NVLTLEQRKF YEENGFLVIK NLVPDADIQR FRNEFEKICR KEVKPLGLTV MRDVTISKSE YAPSEKMITK VQDFQEDKEL FRYCTLPEIL KYVECFTGPN IMAMHTMLIN KPPDSGKKTS RHPLHQDLHY FPFRPSDLIV CAWTAMEHIS RNNGCLVVLP GTHKGSLKPH DYPKWEGGVN KMFHGIQDYE ENKARVHLVM EKGDTVFFHP LLIHGSGQNK TQGFRKAISC HFASADCHYI DVKGTSQENI EKEVVGIAHK FFGAENSVNL KDIWMFRARL VKGERTNL
PHYH2 catalyzes a carbon-carbon cleavage reaction; cleaves a 2-hydroxy-3-methylacyl-CoA into formyl-CoA and a 2-methyl-branched fatty aldehyde.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1600020H07Rik; HPCL; HPCL2; OTTHUMP00000019131; OTTHUMP00000019132; OTTHUMP00000179083; OTTHUMP00000216226; PhyH; PHYH2; Phytanic acid oxidase; phytanoil-CoA alpha hydroxylase; phytanoyl-CoA 2 oxoglutarate dioxygenase; Phytanoyl-CoA alpha-hydroxylase; Phytanoyl-CoA dioxygenase, peroxisomal
Gene Aliases: LN1; LNAP1; PAHX; PHYH; PHYH1; RD
UniProt ID: (Human) O14832
Entrez Gene ID: (Human) 5264
Molecular Function:
hydroxylase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support