Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MHGSCSFLM LLLPLLLLLV ATTGPVGALT DEEKRLMVEL HNLYRAQVSP TASDMLHMRW DEELAAFAKA YARQCVWGHN KERGRRGENL FAITDEGMDV PLAMEEWHHE REHYNLSAAT CSPGQMCGHY TQVVWAKTER IGCGSHFCEK LQGVEETNIE LLVCNYEPPG NVKGKRPYQE GTPCSQCPSG YHCKNSLCEP IGSPEDAQDL PYLVTEAPSF RATEASDSRK MGTPSSLATG IPAFLVTEVS GSLATKALPA VETQAPTSLA TKDPPSMATE APPCVTTEVP SILAAHSLPS L (1-300 aa encoded by BC022399)
May inhibit cardiomyocyte growth. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD364; CRISP-9; Cysteine-rich secretory protein 9; microseminoprotein, beta-binding protein; Peptidase inhibitor 16; PI-16; protease inhibitor 16; PSP94-binding protein
Gene Aliases: CD364; CRISP9; MSMBBP; PI16; PSEC0164; PSPBP; UNQ289/PRO328
UniProt ID: (Human) Q6UXB8
Entrez Gene ID: (Human) 221476
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support