Product References
Adiponectin receptor agonist AdipoRon modulates human and mouse platelet function.
Acta pharmacologica Sinica
Zhou XH,Cheng ZP,Lu M,Lin WY,Luo LL,Ming ZY,Hu Y
Wed Feb 01 00:00:00 EST 2023
Acacetin inhibits myocardial mitochondrial dysfunction by activating PI3K/AKT in SHR rats fed with fructose.
Journal of natural medicines
Yuan P,Zhang Q,Fu Y,Hou Y,Gao L,Wei Y,Feng W,Zheng X
Wed Mar 01 00:00:00 EST 2023
An Integrative Pharmacology-Based Strategy to Uncover the Mechanism of Zuogui Jiangtang Shuxin Formula in Diabetic Cardiomyopathy.
Drug design, development and therapy
Huang Y,Zhang Y,Wu Y,Xiang Q,Yu R
Fri Feb 03 00:00:00 EST 2023
Inhibitory Effect of Hernandezine on the Proliferation of Hepatocellular Carcinoma.
Biological & pharmaceutical bulletin
Kuok C,Wang Q,Fong P,Qin Y,Meng L
Mon Feb 06 00:00:00 EST 2023
20S-Protopanaxatriol improves cognitive function of Alzheimer's disease by promoting endogenous neurogenesis.
Food & function
Zhang Q,Yang X,Liu J,Song J,Zhang S,Chen L,Zhang M
Thu May 11 00:00:00 EDT 2023
PDK1 inhibition reduces autophagy and cell senescence through the PI3K/AKT signalling pathway in a cigarette smoke mouse emphysema model.
Experimental and therapeutic medicine
Zhang P,Jiang Y,Ye X,Zhang C,Tang Y
Mon May 01 00:00:00 EDT 2023
Gegen Qinlian standard decoction alleviated irinotecan-induced diarrhea via PI3K/AKT/NF-ฮบB axis by network pharmacology prediction and experimental validation combination.
Chinese medicine
Chen J,Li M,Chen R,Xu Z,Yang X,Gu H,Zhang L,Fu C,Zhang J,Wu Y
Thu Apr 27 00:00:00 EDT 2023
Erxian decoction alleviates cisplatin-induced premature ovarian failure in rats by reducing oxidation levels in ovarian granulosa cells.
Journal of ethnopharmacology
Liu J,Yang Y,He Y,Feng C,Ou H,Yang J,Chen Y,You F,Shao B,Bao J,Guan X,Chen F,Zhao P
Sat Mar 25 00:00:00 EDT 2023
Chemical Constituents and Hypoglycemic Mechanisms of Dendrobium nobile in Treatment of Type 2 Diabetic Rats by UPLC-ESI-Q-Orbitrap, Network Pharmacology and In Vivo Experimental Verification.
Molecules (Basel, Switzerland)
Li Z,Zeng M,Geng K,Lai D,Xu Z,Zhou W
Thu Mar 16 00:00:00 EDT 2023
Anti-cervical cancer effects of Compound Yangshe granule through the PI3K/AKT pathway based on network pharmacology.
Journal of ethnopharmacology
Ma C,Gu Y,Liu C,Tang X,Yu J,Li D,Liu J
Mon Jan 30 00:00:00 EST 2023
Liraglutide Attenuates Myocardial Ischemia/Reperfusion Injury Through the Inhibition of Necroptosis by Activating GLP-1R/PI3K/Akt Pathway.
Cardiovascular toxicology
Zhou G,Wu H,Yang J,Ye M,Liu D,Li Y,Zhang D,Zhang J,Yang Q,Liu Y
Sat Apr 01 00:00:00 EDT 2023
Enterococcus faecalis promotes the progression of colorectal cancer via its metabolite: biliverdin.
Journal of translational medicine
Zhang L,Liu J,Deng M,Chen X,Jiang L,Zhang J,Tao L,Yu W,Qiu Y
Thu Feb 02 00:00:00 EST 2023
Cytotoxic Steroidal Saponins Containing a Rare Fructosyl from the Rhizomes of Paris polyphylla var. latifolia.
International journal of molecular sciences
Li TY,Du Y,Wang MC,Liu K,Liu Y,Cao Y,Wang YY,Chen WW,Qian XY,Qiu PC,Tang HF,Lu YY
Wed Apr 12 00:00:00 EDT 2023
The circHMGCS1-miR-205-5p-ErBB3 axis mediated the Sanggenon C-induced anti-proliferation effects on human prostate cancer.
Pharmacological research
Li J,Cao X,Chu T,Lin K,Chen L,Lv J,Tan Y,Chen M,Li M,Wang K,Zheng Q,Li D
Sun Jan 01 00:00:00 EST 2023
Identification of a neutrophil-specific PIK3R1 mutation facilitates targeted treatment in a patient with Sweet syndrome.
The Journal of clinical investigation
Bhattacharya S,Basu S,Sheng E,Murphy C,Wei J,Kersh AE,Nelson CA,Bryer JS,Ashchyan HA,Steele K,Forrestel A,Seykora JT,Micheletti RG,James WD,Rosenbach M,Leung TH
Tue Jan 03 00:00:00 EST 2023
Jia-Wei-Si-Miao-Yong-An Fang stimulates the healing of acute radiation-induced cutaneous wounds through MAPK/ERK pathway.
Journal of ethnopharmacology
Wang Y,Gao J,Sun L,Li Q,Kang N,Gao C,Li T
Mon Apr 24 00:00:00 EDT 2023
Mechanism Repositioning Based on Integrative Pharmacology: Anti-Inflammatory Effect of Safflower in Myocardial Ischemia-Reperfusion Injury.
International journal of molecular sciences
Zhao F,Jiang H,Zhang T,Chen H,Li W,Li X,Wang P,Xu H
Fri Mar 10 00:00:00 EST 2023
ANRIL promotes the regulation of colorectal cancer on lymphatic endothelial cells via VEGF-C and is the key target for Pien Tze Huang to inhibit cancer metastasis.
Cancer gene therapy
Huang B,Lu Y,Ni Z,Liu J,He Y,An H,Ye F,Shen J,Lin M,Chen Y,Lin J
Fri Sep 01 00:00:00 EDT 2023
Network Pharmacology and Experimental Verification to Explore the Potential Mechanism of Yin-Huo-Tang for Lung Adenocarcinoma Recurrence.
Drug design, development and therapy
Liu D,Lin S,Li Y,Zhou T,Hu K,Li Q
Mon Apr 04 00:00:00 EDT 2022
Glytabastan B, a coumestan isolated from Glycine tabacina, alleviated synovial inflammation, osteoclastogenesis and collagen-induced arthritis through inhibiting MAPK and PI3K/AKT pathways.
Biochemical pharmacology
Tu Y,Tan L,Lu T,Wang K,Wang H,Han B,Zhao Y,Chen H,Li Y,Chen H,Chen M,He C
Tue Mar 01 00:00:00 EST 2022
OEA alleviates apoptosis in diabetic rats with myocardial ischemia/reperfusion injury by regulating the PI3K/Akt signaling pathway through activation of TRPV1.
Frontiers in pharmacology
Yao E,Luo L,Lin C,Wen J,Li Y,Ren T,Chen Y,Huang J,Jin X
Fri Dec 02 00:00:00 EST 2022
OEA alleviates apoptosis in diabetic rats with myocardial ischemia/reperfusion injury by regulating the PI3K/Akt signaling pathway through activation of TRPV1.
Frontiers in pharmacology
Yao E,Luo L,Lin C,Wen J,Li Y,Ren T,Chen Y,Huang J,Jin X
Fri Dec 02 00:00:00 EST 2022
Upregulation of MHC-I and downregulation of PD-L1 expression by doxorubicin and deferasirox codelivered liposomal nanoparticles for chemoimmunotherapy of melanoma.
International journal of pharmaceutics
Song P,Han X,Zheng R,Yan J,Wu X,Wang Y,Zhang H
Thu Aug 25 00:00:00 EDT 2022
D-Mannose Regulates Hepatocyte Lipid Metabolism via PI3K/Akt/mTOR Signaling Pathway and Ameliorates Hepatic Steatosis in Alcoholic Liver Disease.
Frontiers in immunology
Hu M,Chen Y,Deng F,Chang B,Luo J,Dong L,Lu X,Zhang Y,Chen Z,Zhou J
Tue Apr 26 00:00:00 EDT 2022
Xihuang pills induce apoptosis in hepatocellular carcinoma by suppressing phosphoinositide 3-kinase/protein kinase-B/mechanistic target of rapamycin pathway.
World journal of gastrointestinal oncology
Teng YJ,Deng Z,Ouyang ZG,Zhou Q,Mei S,Fan XX,Wu YR,Long HP,Fang LY,Yin DL,Zhang BY,Guo YM,Zhu WH,Huang Z,Zheng P,Ning DM,Tian XF
Fri Apr 15 00:00:00 EDT 2022
Hua-Tan-Sheng-Jing Decoction Treats Obesity With Oligoasthenozoospermia by Up-Regulating the PI3K-AKT and Down-Regulating the JNK MAPK Signaling Pathways: At the Crossroad of Obesity and Oligoasthenozoospermia.
Frontiers in pharmacology
Dong Y,Zheng Y,Zhu L,Li T,Guan Y,Zhao S,Wang Q,Wang J,Li L
Sat Jul 16 00:00:00 EDT 2022
lncRNA MPFAST Promotes Proliferation and Fatty Acid Synthesis of Bovine Mammary Epithelial Cell by Sponging miR-103 Regulating PI3K-AKT Pathway.
Journal of agricultural and food chemistry
Liu L,Sun B,Zhang F,Zhong Z,Zhang Y,Li F,Zhang T,Khatib H,Wang X
Wed Sep 28 00:00:00 EDT 2022
Hexokinase 2-mediated glycolysis promotes receptor activator of NF-ฮบB ligand expression in Porphyromonas gingivalis lipopolysaccharide-treated osteoblasts.
Journal of periodontology
Yu Y,Jiang L,Li J,Lei L,Li H
Fri Jul 01 00:00:00 EDT 2022
Glucocorticoids induce osteonecrosis of the femoral head in rats via PI3K/AKT/FOXO1 signaling pathway.
PeerJ
Sun F,Zhou JL,Wei SX,Jiang ZW,Peng H
Fri Jan 13 00:00:00 EST 2023
The Role of Syncytin in Placental Angiogenesis and Fetal Growth.
Frontiers in cell and developmental biology
Wang YN,Ye Y,Zhou D,Guo ZW,Xiong Z,Gong XX,Jiang SW,Chen H
Sat Jul 16 00:00:00 EDT 2022
Rab22a promotes the proliferation, migration, and invasion of lung adenocarcinoma via up-regulating PI3K/Akt/mTOR signaling pathway.
Experimental cell research
Wang J,Luo X,Lu J,Wang X,Miao Y,Li Q,Wang L
Fri Jul 15 00:00:00 EDT 2022
Theabrownins Produced via Chemical Oxidation of Tea Polyphenols Inhibit Human Lung Cancer Cells in vivo and in vitro by Suppressing the PI3K/AKT/mTOR Pathway Activation and Promoting Autophagy.
Frontiers in nutrition
Wang Y,Yuan Y,Wang C,Wang B,Zou W,Zhang N,Chen X
Tue May 10 00:00:00 EDT 2022
Picropodophyllin Inhibits the Proliferation of Human Prostate Cancer DU145 and LNCaP Cells via ROS Production and PI3K/AKT Pathway Inhibition.
Biological & pharmaceutical bulletin
Zhu X,Chen X,Wang G,Lei D,Chen X,Lin K,Li M,Lin H,Li D,Zheng Q
Tue Aug 02 00:00:00 EDT 2022
INPP4B inhibits glioma cell proliferation and immune escape via inhibition of the PI3K/AKT signaling pathway.
Frontiers in oncology
Sun X,Chen Y,Tao X,Zhang W,Wang X,Wang X,Ruan Z,Chen Z
Sat Sep 24 00:00:00 EDT 2022
Combined exposure of lead and high-fat diet enhanced cognitive decline via interacting with CREB-BDNF signaling in male rats.
Environmental pollution (Barking, Essex : 1987)
Liu R,Bai L,Liu M,Wang R,Wu Y,Li Q,Ba Y,Zhang H,Zhou G,Yu F,Huang H
Fri Jul 01 00:00:00 EDT 2022
Anti-PD-L1 antibody alleviates pulmonary fibrosis by inducing autophagy via inhibition of the PI3K/Akt/mTOR pathway.
International immunopharmacology
Lu Y,Zhong W,Liu Y,Chen W,Zhang J,Zeng Z,Huang H,Qiao Y,Wan X,Meng X,Cai S,Dong H
Tue Mar 01 00:00:00 EST 2022
Protective Effect of Photobiomodulation against Hydrogen Peroxide-Induced Oxidative Damage by Promoting Autophagy through Inhibition of PI3K/AKT/mTOR Pathway in MC3T3-E1 Cells.
Oxidative medicine and cellular longevity
Zuo X,Wei X,Ju C,Wang X,Zhang Z,Ma Y,Zhu Z,Li X,Song Z,Luo L,Hu X,Wang Z
Mon Dec 05 00:00:00 EST 2022
Prediction of the Mechanism of Shaoyao Gancao Decoction in the Treatment of Alopecia Areata by Network Pharmacology and Its Preliminary Verification Study.
Evidence-based complementary and alternative medicine : eCAM
Lv S,Wang L,Duan Y,Huang D,Yang D
Sat Jul 16 00:00:00 EDT 2022
Cyclo-(Phe-Tyr) as a novel cyclic dipeptide compound alleviates ischemic/reperfusion brain injury via JUNB/JNK/NF-ฮบB and SOX5/PI3K/AKT pathways.
Pharmacological research
Cai J,Liang J,Zhang Y,Shen L,Lin H,Hu T,Zhan S,Xie M,Liang S,Xian M,Wang S
Wed Jun 01 00:00:00 EDT 2022
Downregulation of NCL attenuates tumor formation and growth in HeLa cells by targeting the PI3K/AKT pathway.
Cancer medicine
Ying J,Pan R,Tang Z,Zhu J,Ren P,Lou Y,Zhang E,Huang D,Hu P,Li D,Bao Q,Li P
Tue Mar 01 00:00:00 EST 2022
Network pharmacology and molecular docking integrated strategy to investigate the pharmacological mechanism of palmatine in Alzheimer's disease.
Journal of biochemical and molecular toxicology
Pei H,Zeng J,Chen W,He Z,Du R
Tue Nov 01 00:00:00 EDT 2022
Dopamine receptor D2 regulates GLUA1-containing AMPA receptor trafficking and central sensitization through the PI3K signaling pathway in a male rat model of chronic migraine.
The journal of headache and pain
Zhang W,Lei M,Wen Q,Zhang D,Qin G,Zhou J,Chen L
Wed Aug 10 00:00:00 EDT 2022
Honokiol improves depression-like behaviors in rats by HIF-1ฮฑ- VEGF signaling pathway activation.
Frontiers in pharmacology
Fan XX,Sun WY,Li Y,Tang Q,Li LN,Yu X,Wang SY,Fan AR,Xu XQ,Chang HS
Tue Sep 13 00:00:00 EDT 2022
Fu Brick Tea Manages HFD/STZ-Induced Type 2 Diabetes by Regulating the Gut Microbiota and Activating the IRS1/PI3K/Akt Signaling Pathway.
Journal of agricultural and food chemistry
Qi B,Ren D,Li T,Niu P,Zhang X,Yang X,Xiao J
Wed Jul 13 00:00:00 EDT 2022
High yield engineered nanovesicles from ADSC with enriched miR-21-5p promote angiogenesis in adipose tissue regeneration.
Biomaterials research
Sun D,Mou S,Chen L,Yang J,Wang R,Zhong A,Wang W,Tong J,Wang Z,Sun J
Sat Dec 17 00:00:00 EST 2022
Exploring the pharmacological mechanism of calculus bovis in cerebral ischaemic stroke using a network pharmacology approach.
Journal of ethnopharmacology
Du X,Li C,Zhang S,Sun C,Zhang X,Chen C,Wang X,Cheng F,Wang Q
Thu Feb 10 00:00:00 EST 2022
Network Pharmacology-Based Prediction and Verification of the Potential Targets of Pinoresinol Diglucoside for OA Treatment.
Evidence-based complementary and alternative medicine : eCAM
Lou H,Zhang Y,Fang J,Jin Y
Sat Jul 16 00:00:00 EDT 2022
Fucoidan ameliorates glucose metabolism by the improvement of intestinal barrier and inflammatory damage in type 2 diabetic rats.
International journal of biological macromolecules
Liu Y,Xu Z,Huang H,Xue Y,Zhang D,Zhang Y,Li W,Li X
Tue Mar 15 00:00:00 EDT 2022
Fucoidan protects the pancreas and improves glucose metabolism through inhibiting inflammation and endoplasmic reticulum stress in T2DM rats.
Food & function
Liu Y,Huang H,Xu Z,Xue Y,Zhang D,Zhang Y,Li W,Li X
Mon Mar 07 00:00:00 EST 2022
Triclosan regulates the Nrf2/HO-1 pathway through the PI3K/Akt/JNK signaling cascade to induce oxidative damage in neurons.
Environmental toxicology
Wang D,Liu J,Jiang H
Wed Sep 01 00:00:00 EDT 2021
Kaempferol-induced GPER upregulation attenuates atherosclerosis via the PI3K/AKT/Nrf2 pathway.
Pharmaceutical biology
Feng Z,Wang C,Meng Q,Wu J,Sun H
Wed Dec 01 00:00:00 EST 2021
Qingjie Fuzheng Granule suppresses lymphangiogenesis in colorectal cancer via the VEGF-C/VEGFR-3 dependent PI3K/AKT pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Huang B,Lu Y,Gui M,Guan J,Lin M,Zhao J,Mao Q,Lin J
Sat May 01 00:00:00 EDT 2021
MiR-19a/miR-96-mediated low expression of KIF26A suppresses metastasis by regulating FAK pathway in gastric cancer.
Oncogene
Ma RR,Zhang H,Chen HF,Zhang GH,Tian YR,Gao P
Thu Apr 01 00:00:00 EDT 2021
Xiaoyaosan Alleviates Hippocampal Glutamate-Induced Toxicity in the CUMS Rats via NR2B and PI3K/Akt Signaling Pathway.
Frontiers in pharmacology
Zhou XM,Liu CY,Liu YY,Ma QY,Zhao X,Jiang YM,Li XJ,Chen JX
Mon Jan 02 00:00:00 EST 2023
The protein kinase D1-mediated inflammatory pathway is involved in olanzapine-induced impairment of skeletal muscle insulin signaling in rats.
Life sciences
Wang C,Wang C,Ren L,Chen S,Chen WH,Li Y
Thu Apr 01 00:00:00 EDT 2021
ABCC5 facilitates the acquired resistance of sorafenib through the inhibition of SLC7A11-induced ferroptosis in hepatocellular carcinoma.
Neoplasia (New York, N.Y.)
Huang W,Chen K,Lu Y,Zhang D,Cheng Y,Li L,Huang W,He G,Liao H,Cai L,Tang Y,Zhao L,Pan M
Wed Dec 01 00:00:00 EST 2021
Clozapine Induced Disturbances in Hepatic Glucose Metabolism: The Potential Role of PGRMC1 Signaling.
Frontiers in endocrinology
Cao T,Chen Q,Zhang B,Wu X,Zeng C,Zhang S,Cai H
Mon Feb 14 00:00:00 EST 2022
Chitosan oligosaccharides packaged into rat adipose mesenchymal stem cells-derived extracellular vesicles facilitating cartilage injury repair and alleviating osteoarthritis.
Journal of nanobiotechnology
Li S,Liu J,Liu S,Jiao W,Wang X
Tue Oct 26 00:00:00 EDT 2021
Morphine promotes theย malignant biological behavior of non-small cell lung cancer cells through the MOR/Src/mTOR pathway.
Cancer cell international
Liu X,Yang J,Yang C,Huang X,Han M,Kang F,Li J
Thu Nov 25 00:00:00 EST 2021
Daphnoretin induces reactive oxygen species-mediated apoptosis in melanoma cells.
Oncology letters
Wang H,Hu X,Li M,Pan Z,Li D,Zheng Q
Tue Jun 01 00:00:00 EDT 2021
Aloe-emodin prevents nerve injury and neuroinflammation caused by ischemic stroke via the PI3K/AKT/mTOR and NF-ฮบB pathway.
Food & function
Xian M,Cai J,Zheng K,Liu Q,Liu Y,Lin H,Liang S,Wang S
Tue Sep 07 00:00:00 EDT 2021
Aronia melanocarpa polysaccharide ameliorates inflammation and aging in mice by modulating the AMPK/SIRT1/NF-ฮบB signaling pathway and gut microbiota.
Scientific reports
Zhao Y,Liu X,Zheng Y,Liu W,Ding C
Mon Oct 18 00:00:00 EDT 2021
Luteolin Alleviates Methamphetamine-Induced Hepatotoxicity by Suppressing the p53 Pathway-Mediated Apoptosis, Autophagy, and Inflammation in Rats.
Frontiers in pharmacology
Zhang KK,Wang H,Qu D,Chen LJ,Wang LB,Li JH,Liu JL,Xu LL,Yoshida JS,Xu JT,Xie XL,Li DR
Wed Mar 10 00:00:00 EST 2021
Fatty Acid-Binding Protein 4 (FABP4) Suppresses Proliferation and Migration of Endometrial Cancer Cells via PI3K/Akt Pathway.
OncoTargets and therapy
Wu Z,Jeong JH,Ren C,Yang L,Ding L,Li F,Jiang D,Zhu Y,Lu J
Sun Apr 24 00:00:00 EDT 2022
Ganoderma lucidum Spore Polysaccharide Inhibits the Growth of Hepatocellular Carcinoma Cells by Altering Macrophage Polarity and Induction of Apoptosis.
Journal of immunology research
Song M,Li ZH,Gu HS,Tang RY,Zhang R,Zhu YL,Liu JL,Zhang JJ,Wang LY
Thu Nov 11 00:00:00 EST 2021
miR-126 Controls the Apoptosis and Proliferation of Immature Porcine Sertoli Cells by Targeting the PIK3R2 Gene through the PI3K/AKT Signaling Pathway.
Animals : an open access journal from MDPI
Tang X,Chen Y,Luo H,Bian Q,Weng B,Yang A,Chu D,Ran M,Chen B
Fri Jul 30 00:00:00 EDT 2021
Exploring the Pharmacological Mechanisms of Xihuang Pills Against Prostate Cancer via Integrating Network Pharmacology and Experimental Validation In Vitro and In Vivo.
Frontiers in pharmacology
Wu Y,You X,Lin Q,Xiong W,Guo Y,Huang Z,Dai X,Chen Z,Mei S,Long Y,Tian X,Zhou Q
Tue Mar 29 00:00:00 EDT 2022
B7-H3 inhibits apoptosis of gastric cancer cell by interacting with Fibronectin.
Journal of Cancer
Sun M,Xie J,Zhang D,Chen C,Lin S,Chen Y,Zhang G
Sat Feb 19 00:00:00 EST 2022
Parthenolide inhibits the tumor characteristics of renal cell carcinoma.
International journal of oncology
Liu D,Han Y,Liu L,Ren X,Zhang H,Fan S,Qin T,Li L
Fri Jan 01 00:00:00 EST 2021
Downregulation of GLYAT Facilitates Tumor Growth and Metastasis and Poor Clinical Outcomes Through the PI3K/AKT/Snail Pathway in Human Breast Cancer.
Frontiers in oncology
Tian X,Wu L,Jiang M,Zhang Z,Wu R,Miao J,Liu C,Gao S
Sat Jul 16 00:00:00 EDT 2022
Babao Dan Reverses Multiple-Drug Resistance in Gastric Cancer Cells via Triggering Apoptosis and Autophagy and Inhibiting PI3K/AKT/mTOR Signaling.
Evidence-based complementary and alternative medicine : eCAM
Zhao J,Lan W,Peng J,Guan B,Liu J,Zhang M,Zhan Z,Lin J
Tue Jul 27 00:00:00 EDT 2021
A Potential Mechanism Underlying the Therapeutic Effects of Progesterone and Allopregnanolone on Ketamine-Induced Cognitive Deficits.
Frontiers in pharmacology
Cao T,Tang M,Jiang P,Zhang B,Wu X,Chen Q,Zeng C,Li N,Zhang S,Cai H
Sat Mar 27 00:00:00 EDT 2021
CHRDL2 promotes osteosarcoma cell proliferation and metastasis through the BMP-9/PI3K/AKT pathway.
Cell biology international
Chen H,Pan R,Li H,Zhang W,Ren C,Lu Q,Chen H,Zhang X,Nie Y
Mon Mar 01 00:00:00 EST 2021
Galectin-3 mediates cardiac remodeling caused by impaired glucose and lipid metabolism through inhibiting two pathways of activating Akt.
American journal of physiology. Heart and circulatory physiology
Sun Z,Zhang L,Li L,Shao C,Liu J,Zhou M,Wang Z
Fri Jan 01 00:00:00 EST 2021
Characteristics of inflammation process in monocrotaline-induced pulmonary arterial hypertension in rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Tang C,Luo Y,Li S,Huang B,Xu S,Li L
Fri Jan 01 00:00:00 EST 2021
The P2RY12 receptor promotes VSMC-derived foam cell formation by inhibiting autophagy in advanced atherosclerosis.
Autophagy
Pi S,Mao L,Chen J,Shi H,Liu Y,Guo X,Li Y,Zhou L,He H,Yu C,Liu J,Dang Y,Xia Y,He Q,Jin H,Li Y,Hu Y,Miao Y,Yue Z,Hu B
Thu Apr 01 00:00:00 EDT 2021
Effects of total flavonoids from Eucommia ulmoides Oliv. leaves on polycystic ovary syndrome with insulin resistance model rats induced by letrozole combined with a high-fat diet.
Journal of ethnopharmacology
Peng MF,Tian S,Song YG,Li CX,Miao MS,Ren Z,Li M
Sat Jun 12 00:00:00 EDT 2021
Photobiomodulation Promotes Repair Following Spinal Cord Injury by Regulating the Transformation of A1/A2 Reactive Astrocytes.
Frontiers in neuroscience
Wang X,Zhang Z,Zhu Z,Liang Z,Zuo X,Ju C,Song Z,Li X,Hu X,Wang Z
Sat Nov 20 00:00:00 EST 2021
Targeting IFN-ฮณ-inducible lysosomal thiol reductase overcomes chemoresistance in AML through regulating the ROS-mediated mitochondrial damage.
Translational oncology
Niu LT,Wang YQ,Wong CCL,Gao SX,Mo XD,Huang XJ
Wed Sep 01 00:00:00 EDT 2021
lncRNA Gm16410 Mediates PM2.5-Induced Macrophage Activation via PI3K/AKT Pathway.
Frontiers in cell and developmental biology
Xu J,Xu H,Ma K,Wang Y,Niu B,Zhang L,Li F
Sat Apr 03 00:00:00 EDT 2021
Exendin-4, a GLP-1 receptor agonist regulates retinal capillary tone and restores microvascular patency after ischaemia-reperfusion injury.
British journal of pharmacology
Zhai R,Xu H,Hu F,Wu J,Kong X,Sun X
Sat Aug 01 00:00:00 EDT 2020
Hepatitis B virus upregulates cellular inhibitor of apoptosis protein 2 expression via the PI3K/AKT/NF-ฮบB signaling pathway in liver cancer.
Oncology letters
Lian J,Zou Y,Huang L,Cheng H,Huang K,Zeng J,Chen L
Sun Mar 01 00:00:00 EST 2020
Endosulfan promotes cell migration via PTP4A3-mediated signaling pathways in HUVECs.
Ecotoxicology and environmental safety
Li H,Liu S,Hu Y,Zhao B,Sun Y,Xu D
Wed Apr 01 00:00:00 EDT 2020
miRNA-98-5p Targeting IGF2BP1 Induces Mesenchymal Stem Cell Apoptosis by Modulating PI3K/Akt and p53 in Immune Thrombocytopenia.
Molecular therapy. Nucleic acids
Wang Y,Zhang J,Su Y,Wang C,Zhang G,Liu X,Chen Q,Lv M,Chang Y,Peng J,Hou M,Huang X,Zhang X
Fri Jun 05 00:00:00 EDT 2020
Involvement of CXCL12/CXCR4 in the motility of human first-trimester endometrial epithelial cells through an autocrine mechanism by activating PI3K/AKT signaling.
BMC pregnancy and childbirth
Zheng J,Qu D,Wang C,Ding L,Zhou W
Mon Feb 10 00:00:00 EST 2020
Naringin alleviates H2O2-induced apoptosis via the PI3K/Akt pathway in rat nucleus pulposus-derived mesenchymal stem cells.
Connective tissue research
Nan LP,Wang F,Ran D,Zhou SF,Liu Y,Zhang Z,Huang ZN,Wang ZY,Wang JC,Feng XM,Zhang L
Sun Nov 01 00:00:00 EDT 2020
Disordered Leptin signaling in the retrotrapezoid nucleus is associated with the impaired hypercapnic ventilatory response in obesity.
Life sciences
Wei Z,Hao Y,Yu H,Shi L,Jing X,Zhang X,Liu N,Li T,Zhang X,Yuan F
Tue Sep 15 00:00:00 EDT 2020
CAPN1 promotes malignant behavior and erlotinib resistance mediated by phosphorylation of c-Met and PIK3R2 via degrading PTPN1 in lung adenocarcinoma.
Thoracic cancer
Chen Y,Tang J,Lu T,Liu F
Wed Jul 01 00:00:00 EDT 2020
Exogenous Therapeutics of Microrna-29a Attenuates Development of Hepatic Fibrosis in Cholestatic Animal Model through Regulation of Phosphoinositide 3-Kinase p85 Alpha.
International journal of molecular sciences
Yang YL,Wang FS,Lin HY,Huang YH
Thu May 21 00:00:00 EDT 2020
Short-Term Exposure to Waterpipe/Hookah Smoke Triggers a Hyperactive Platelet Activation State and Increases the Risk of Thrombogenesis.
Arteriosclerosis, thrombosis, and vascular biology
Alarabi AB,Karim ZA,Ramirez JEM,Hernandez KR,Lozano PA,Rivera JO,Alshbool FZ,Khasawneh FT
Sat Feb 01 00:00:00 EST 2020
HPV16 E6/E7 promote the translocation and glucose uptake of GLUT1 by PI3K/AKT pathway via relieving miR-451 inhibitory effect on CAB39 in lung cancer cells.
Therapeutic advances in chronic disease
Wang HM,Lu YJ,He L,Gu NJ,Wang SY,Qiu XS,Wang EH,Wu GP
Sun Apr 17 00:00:00 EDT 2022
Eurocristatine, a plant alkaloid from Eurotium cristatum, alleviates insulin resistance in db/db diabetic mice via activation of PI3K/AKT signaling pathway.
European journal of pharmacology
Zhang H,Hui J,Yang J,Deng J,Fan D
Sun Nov 15 00:00:00 EST 2020
Eurocristatine, a plant alkaloid from Eurotium cristatum, alleviates insulin resistance in db/db diabetic mice via activation of PI3K/AKT signaling pathway.
European journal of pharmacology
Zhang H,Hui J,Yang J,Deng J,Fan D
Sun Nov 15 00:00:00 EST 2020
Eurocristatine, a plant alkaloid from Eurotium cristatum, alleviates insulin resistance in db/db diabetic mice via activation of PI3K/AKT signaling pathway.
European journal of pharmacology
Zhang H,Hui J,Yang J,Deng J,Fan D
Sun Nov 15 00:00:00 EST 2020
Eurocristatine, a plant alkaloid from Eurotium cristatum, alleviates insulin resistance in db/db diabetic mice via activation of PI3K/AKT signaling pathway.
European journal of pharmacology
Zhang H,Hui J,Yang J,Deng J,Fan D
Sun Nov 15 00:00:00 EST 2020
Inhibition of Specificity Protein 1 Is Involved in Phloretin-Induced Suppression of Prostate Cancer.
BioMed research international
Kang D,Zuo W,Wu Q,Zhu Q,Liu P
Thu Apr 29 00:00:00 EDT 2021
QiShenYiQi pill improves the reparative myocardial fibrosis by regulating autophagy.
Journal of cellular and molecular medicine
Lv S,Yuan P,Dong J,Lu C,Li M,Qu F,Zhu Y,Yuan Z,Zhang J
Thu Oct 01 00:00:00 EDT 2020
Microglia induce the transformation of A1/A2 reactive astrocytes via the CXCR7/PI3K/Akt pathway in chronic post-surgical pain.
Journal of neuroinflammation
Li T,Liu T,Chen X,Li L,Feng M,Zhang Y,Wan L,Zhang C,Yao W
Tue Jul 14 00:00:00 EDT 2020
MiR-155 promotes interleukin-1ฮฒ-induced chondrocyte apoptosis and catabolic activity by targeting PIK3R1-mediated PI3K/Akt pathway.
Journal of cellular and molecular medicine
Fan Z,Liu Y,Shi Z,Deng K,Zhang H,Li Q,Cao S,Li S,Zhang H
Sat Aug 01 00:00:00 EDT 2020
Protective effect of astaxanthin against La2O3 nanoparticles induced neurotoxicity by activating PI3K/AKT/Nrf-2 signaling in mice.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Yuan L,Qu Y,Li Q,An T,Chen Z,Chen Y,Deng X,Bai D
Thu Oct 01 00:00:00 EDT 2020
Glutathione S-transferases P1 protects breast cancer cell from adriamycin-induced cell death through promoting autophagy.
Cell death and differentiation
Dong X,Yang Y,Zhou Y,Bi X,Zhao N,Zhang Z,Li L,Hang Q,Zhang R,Chen D,Cao P,Yin Z,Luo L
Tue Oct 01 00:00:00 EDT 2019
Exosomes Play an Important Role in the Progression of Plasma Cell Mastitis via the PI3K-Akt-mTOR Signaling Pathway.
Mediators of inflammation
Wang X,Han Y,Liu J,Zhang Y,Cheng K,Guo J,Guo Q,Liu S,Sun H,Hua Y,Zhang G,Xu S,Guo F,Yang Z
Mon Jan 13 00:00:00 EST 2020
Exosomes Play an Important Role in the Progression of Plasma Cell Mastitis via the PI3K-Akt-mTOR Signaling Pathway.
Mediators of inflammation
Wang X,Han Y,Liu J,Zhang Y,Cheng K,Guo J,Guo Q,Liu S,Sun H,Hua Y,Zhang G,Xu S,Guo F,Yang Z
Mon Jan 13 00:00:00 EST 2020
TNF-ฮฑ Regulates ITGฮฒ1 and SYND4 Expression in Nucleus Pulposus Cells: Activation of FAK/PI3K Signaling.
Inflammation
Wu X,Li S,Wang K,Hua W,Li S,Song Y,Zhang Y,Yang S,Yang C
Tue Oct 01 00:00:00 EDT 2019
A Peptide Analogue of Selectin Ligands Attenuated Atherosclerosis by Inhibiting Monocyte Activation.
Mediators of inflammation
Ye Z,Zhang S,Liu Y,Wang S,Zhang J,Huang R
Mon Dec 16 00:00:00 EST 2019
Hyperinsulinemia Can Cause Kidney Disease in the IGT Stage of OLETF Rats via the INS/IRS-1/PI3-K/Akt Signaling Pathway.
Journal of diabetes research
Zhang Y,Yang S,Cui X,Yang J,Zheng M,Jia J,Han F,Yang X,Wang J,Guo Z,Chang B,Chang B
Tue Apr 21 00:00:00 EDT 2020
Minimally modified low-density lipoprotein upregulates the ETB and ฮฑ1 receptors in mouse mesenteric arteries inย vivo by activating the PI3K/Akt pathway.
The Journal of pharmacy and pharmacology
Zeng ZS,Lin J,Xu CB,Cao L,Chen C,Li J
Sat Jun 01 00:00:00 EDT 2019
Tetrandrine-Induced Autophagy in MDA-MB-231 Triple-Negative Breast Cancer Cell through the Inhibition of PI3K/AKT/mTOR Signaling.
Evidence-based complementary and alternative medicine : eCAM
Guo Y,Pei X
Sun Apr 10 00:00:00 EDT 2022
Catalpol induces cell activity to promote axonal regeneration via the PI3K/AKT/mTOR pathway in vivo and in vitro stroke model.
Annals of translational medicine
Wang J,Wan D,Wan G,Wang J,Zhang J,Zhu H
Sun Dec 01 00:00:00 EST 2019
Anticancer effect of SZC015 on pancreatic cancer via mitochondria-dependent apoptosis and the constitutive suppression of activated nuclear factor ฮบB and STAT3 in vitro and in vivo.
Journal of cellular physiology
Song Y,Gao L,Tang Z,Li H,Sun B,Chu P,Qaed E,Ma X,Peng J,Wang S,Hu M,Tang Z
Mon Jan 01 00:00:00 EST 2018
Anticancer effect of SZC015 on pancreatic cancer via mitochondria-dependent apoptosis and the constitutive suppression of activated nuclear factor ฮบB and STAT3 in vitro and in vivo.
Journal of cellular physiology
Song Y,Gao L,Tang Z,Li H,Sun B,Chu P,Qaed E,Ma X,Peng J,Wang S,Hu M,Tang Z
Mon Jan 01 00:00:00 EST 2018
The Antiapoptosis Effect of Geum japonicum Thunb. var. chinense Extracts on Cerebral Ischemia Reperfusion Injury via PI3K/Akt Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ou B,Tao W,Yang S,Feng J,Wang J,Yang T,Wu H,Huang Y,Tan L,Huang W,Feng Z,Mei Z
Wed Sep 30 00:00:00 EDT 2020
The Antiapoptosis Effect of Geum japonicum Thunb. var. chinense Extracts on Cerebral Ischemia Reperfusion Injury via PI3K/Akt Pathway.
Evidence-based complementary and alternative medicine : eCAM
Ou B,Tao W,Yang S,Feng J,Wang J,Yang T,Wu H,Huang Y,Tan L,Huang W,Feng Z,Mei Z
Wed Sep 30 00:00:00 EDT 2020
Traditional Chinese Medicine CFF-1 induced cell growth inhibition, autophagy, and apoptosis via inhibiting EGFR-related pathways in prostate cancer.
Cancer medicine
Wu Z,Zhu Q,Yin Y,Kang D,Cao R,Tian Q,Zhang Y,Lu S,Liu P
Sun Apr 01 00:00:00 EDT 2018
Traditional Chinese Medicine CFF-1 induced cell growth inhibition, autophagy, and apoptosis via inhibiting EGFR-related pathways in prostate cancer.
Cancer medicine
Wu Z,Zhu Q,Yin Y,Kang D,Cao R,Tian Q,Zhang Y,Lu S,Liu P
Sun Apr 01 00:00:00 EDT 2018
Anti-inflammatory effects of curcumin are associated with down regulating microRNA-155 in LPS-treated macrophages and mice.
Pharmaceutical biology
Ma F,Liu F,Ding L,You M,Yue H,Zhou Y,Hou Y
Fri Dec 01 00:00:00 EST 2017
Anti-inflammatory effects of curcumin are associated with down regulating microRNA-155 in LPS-treated macrophages and mice.
Pharmaceutical biology
Ma F,Liu F,Ding L,You M,Yue H,Zhou Y,Hou Y
Fri Dec 01 00:00:00 EST 2017
The role of sodium hydrosulfide in attenuating the aging process via PI3K/AKT and CaMKKฮฒ/AMPK pathways.
Redox biology
Chen X,Zhao X,Cai H,Sun H,Hu Y,Huang X,Kong W,Kong W
Tue Aug 01 00:00:00 EDT 2017
The role of sodium hydrosulfide in attenuating the aging process via PI3K/AKT and CaMKKฮฒ/AMPK pathways.
Redox biology
Chen X,Zhao X,Cai H,Sun H,Hu Y,Huang X,Kong W,Kong W
Tue Aug 01 00:00:00 EDT 2017
CAPE-pNO2 ameliorated diabetic nephropathy through regulating the Akt/NF-ฮบB/ iNOS pathway in STZ-induced diabetic mice.
Oncotarget
Wang X,Li D,Fan L,Xiao Q,Zuo H,Li Z
Fri Dec 29 00:00:00 EST 2017
CAPE-pNO2 ameliorated diabetic nephropathy through regulating the Akt/NF-ฮบB/ iNOS pathway in STZ-induced diabetic mice.
Oncotarget
Wang X,Li D,Fan L,Xiao Q,Zuo H,Li Z
Fri Dec 29 00:00:00 EST 2017
cfa-miR-143 Promotes Apoptosis via the p53 Pathway in Canine Influenza Virus H3N2-Infected Cells.
Viruses
Zhou P,Tu L,Lin X,Hao X,Zheng Q,Zeng W,Zhang X,Zheng Y,Wang L,Li S
Sat Nov 25 00:00:00 EST 2017
cfa-miR-143 Promotes Apoptosis via the p53 Pathway in Canine Influenza Virus H3N2-Infected Cells.
Viruses
Zhou P,Tu L,Lin X,Hao X,Zheng Q,Zeng W,Zhang X,Zheng Y,Wang L,Li S
Sat Nov 25 00:00:00 EST 2017
Hydrogen Sulphide Treatment Increases Insulin Sensitivity and Improves Oxidant Metabolism through the CaMKKbeta-AMPK Pathway in PA-Induced IR C2C12 Cells.
Scientific reports
Chen X,Zhao X,Lan F,Zhou T,Cai H,Sun H,Kong W,Kong W
Mon Oct 16 00:00:00 EDT 2017
Hydrogen Sulphide Treatment Increases Insulin Sensitivity and Improves Oxidant Metabolism through the CaMKKbeta-AMPK Pathway in PA-Induced IR C2C12 Cells.
Scientific reports
Chen X,Zhao X,Lan F,Zhou T,Cai H,Sun H,Kong W,Kong W
Mon Oct 16 00:00:00 EDT 2017
MiRNA-21 induces epithelial to mesenchymal transition and gemcitabine resistance via the PTEN/AKT pathway in breast cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Wu ZH,Tao ZH,Zhang J,Li T,Ni C,Xie J,Zhang JF,Hu XC
Wed Jun 01 00:00:00 EDT 2016
MiRNA-21 induces epithelial to mesenchymal transition and gemcitabine resistance via the PTEN/AKT pathway in breast cancer.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine
Wu ZH,Tao ZH,Zhang J,Li T,Ni C,Xie J,Zhang JF,Hu XC
Wed Jun 01 00:00:00 EDT 2016