Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • PI4K2A Antibodies

            Proteintech

            PI4K2A Polyclonal Antibody

            3 References
            View all (7) PI4K2A antibodies
            Datasheet
            Protocols
            Questions & Answers
            Promotion Available
            Learn more
            Catalog # 15318-1-AP
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (USD)
            492.00
            150 µL
             
            Check your price
            Request bulk or custom format
            Join Aspire™ for free antibodies
            Datasheet
            Protocols
            Questions & Answers

            Cite PI4K2A Polyclonal Antibody

            • Antibody Testing Data (3)
            Application
            PI4K2A Antibody in Western Blot (WB)
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            PI4K2A Antibody (15318-1-AP) in WB

            HepG2 cells were subjected to SDS PAGE followed by western blot with (Product # 15318-1-AP) (PI4K2A antibody) at dilution of 1:800 incubated at room temperature for 1.5 hours.
            View Product

            PI4K2A Antibody in Western Blot (WB)
            PI4K2A Antibody in Western Blot (WB)
            PI4K2A Antibody in Western Blot (WB)

            Product Details

            15318-1-AP

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:500-1:1,000
            View 2 publications 2 publications

            Immunocytochemistry (ICC/IF)

            -
            View 1 publication 1 publication
            Product Specifications

            Species Reactivity

            Human, Mouse, Rat

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            PI4K2A Fusion Protein Ag7564 (131-479 aa encoded by BC003167)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.13 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            PBS, pH 7.3, with 50% glycerol

            Contains

            0.02% sodium azide

            Storage conditions

            -20°C

            Shipping conditions

            Wet ice (domestic); Dry ice (international)

            Product Specific Information

            Immunogen sequence: YQGSSGSYF VKDPQGRIIA VFKPKNEEPY GHLNPKWTKW LQKLCCPCCF GRDCLVLNQG YLSEAGASLV DQKLELNIVP RTKVVYLASE TFNYSAIDRV KSRGKRLALE KVPKVGQRFN RIGLPPKVGS FQLFVEGYKD ADYWLRRFEA EPLPENTNRQ LLLQFERLVV LDYIIRNTDR GNDNWLIKYD CPMDSSSSRD TDWVVVKEPV IKVAAIDNGL AFPLKHPDSW RAYPFYWAWL PQAKVPFSQE IKDLILPKIS DPNFVKDLEE DLYELFKKDP GFDRGQFHKQ IAVMRGQILN LTQALKDNKS PLHLVQMPPV IVETARSHQR SSSESYTQSF QSRKPFFSWW (131-479 aa encoded by BC003167)

            Target Information

            PI4K2A (PI4K2 alpha) lipid kinase or phosphatidylinositol 4-kinase type 2 alpha is involved in the formation of phosphatidylinositolpolyphosphates (PtdInsPs) which are centrally involved in many biologic processes ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4K2A phosphorylates PtdIns at the D-4 position which is an essential step in the biosynthesis of PtdInsPs. PI4K2A is a novel regulator of tumor growth by its action on angiogenesis and HIF-1alpha regulation. PI4K2A generates PtdIns4P-rich domains within the Golgi which regulate targeting of clathrin adaptor AP-1 complexes. PI4K2A is also required for Wnt to regulate LRP6 phosphorylation.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: 55 kDa type II phosphatidylinositol 4-kinase; null; phosphatidylinositol 4-kinase; Phosphatidylinositol 4-kinase type 2-alpha; phosphatidylinositol 4-kinase type II; phosphatidylinositol 4-kinase type II (PI4KII); Phosphatidylinositol 4-kinase type II-alpha

            View more View less

            Gene Aliases: Pi4k2; PI4K2A; PI4KII; PIK42A

            View more View less

            UniProt ID: (Human) Q9BTU6, (Mouse) Q2TBE6, (Rat) Q99M64

            View more View less

            Entrez Gene ID: (Human) 55361, (Mouse) 84095, (Rat) 114554

            View more View less

            Function(s)

            magnesium ion binding 1-phosphatidylinositol 4-kinase activity protein binding ATP binding AP-3 adaptor complex binding nucleotide binding kinase activity transferase activity phosphotransferase activity, alcohol group as acceptor protein complex binding kinase

            Process(es)

            basophil degranulation phosphatidylinositol biosynthetic process phosphatidylinositol phosphorylation mitophagy in response to mitochondrial depolarization phosphorylation
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            united states flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-vj67q:80/100.66.73.9:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0