Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: Jurkat, Raji, K562, THP-1, HL-60, NCI-H929, NIH-3T3, JC, BCL-1.
Predicted reactivity: Mouse (95%), Rat (94%), Chicken (88%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein involved in the regulation of transcription factors involved in MAP kinase signaling. The symbol MIZ1 has also been associated with ZBTB17 which is a different gene located on chromosome 1. Two alternatively spliced transcripts encoding different isoforms have been described.