Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Samples: Mouse heart; Cellular Location: Cytoplasm, Nucleus, centriolar satellite, centrosome, cytoskeleton, microtubule organizing center
Immunogen sequence: MSRKISKESK KVNISSSLES EDISLETTVP TDDISSSEER EGKVRITRQL IERKELLHNI QLLKIELSQK TMMIDNLKVD YLTKIEELEE KLNDALHQKQ
Mediator of progesterone that by acting on the phospholipase A2 enzyme interferes with arachidonic acid metabolism, induces a Th2 biased immune response, and by controlling NK activity exerts an anti-abortive effect.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Centrosomal protein of 90 kDa; CEP90; PIBF; progesterone immunomodulatory binding factor 1; progesterone-induced blocking factor 1; Progesterone-induced-blocking factor 1; RP11-505F3.1
Gene Aliases: C13orf24; CEP90; PIBF; PIBF1; RGD1305077
UniProt ID: (Human) Q8WXW3
Entrez Gene ID: (Human) 10464, (Mouse) 52023, (Rat) 306104
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support