Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRRPLSKCG MEPGGGDASL TLHGLQNRSH GKIKLRKRKS TLYFNTQEKS ARRRGDLLGE NIYLLLFTLA LRILNCFLVQ TSFVPDEYWQ SLEVSHHMVF NYGYLTWEWT ERLRSYTYPL IFASIYKILH LLGKDSVQLL IWIPRLAQAL LSAVADVRLY SLMKQLENQE VARWVFFCQL CSWFTWYCCT RTLTNTMETV LTIIALFYYP LEGSKSMNSV KYSSLVALAF IIRPTAVILW TPLLFRHFCQ EPRKLDLILH HFLPVGFVTL SLSLMIDRIF FGQWTLVQFN FLKFNVLQNL GTFYGSHPWH WYFSQGFPVI LGTHLPFFIH GCYLAP (1-335 aa encoded by B C017711)
Mannosyltransferase involved in glycosylphosphatidylinositol-anchor biosynthesis. Transfers the third alpha-1, 2-mannose to Man2-GlcN-acyl-PI during GPI precursor assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 3; GPI mannosyltransferase III; GPI-MT-III; involved in GPI-anchor biosynthesis; phosphatidylinositol glycan anchor biosynthesis, class B; phosphatidylinositol glycan, class B; Phosphatidylinositol-glycan biosynthesis class B protein; PIG-B
Gene Aliases: GPI-MT-III; PIG-B; PIGB
UniProt ID: (Human) Q92521, (Mouse) Q9JJQ0
Entrez Gene ID: (Human) 9488, (Mouse) 55981, (Rat) 315807
Molecular Function:
glycosyltransferase
transferase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support