Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEDERSFSDI CGGRLALQRR YYSPSCREFC LSCPRLSLRS LTAVTCTVWL AAYGLFTLCE NSMILSAAIF ITLLGLLGYL HFVKIDQETL LIIDSLGIQM TSSYASGKES TTFIEMGKVK DIVINEAIYM QKVIYYLCIL LKDPVEPHGI SQVVPVFQSA KPRLDCLIEV YRSCQEILAH QKATSTSP
This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. The protein encoded by this gene is a subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mrp64; Phosphatidylinositol N-acetylglucosaminyltransferase subunit H; Phosphatidylinositol-glycan biosynthesis class H protein; phosphatidylinositol-glycan biosynthesis, class H protein; PIG-H
Gene Aliases: GPI-H; PIGH
UniProt ID: (Human) Q14442
Entrez Gene ID: (Human) 5283
Molecular Function:
glycosyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support