Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VAVAGLTFF ALSFGFYYEY GWEFLEHTYF YHLTRRDIRH NFSPYFYMLY LTAESKWSFS LGIAAFLPQL (231-299 aa encoded by BC019865)
PIGM encodes a transmembrane protein that is located in the endoplasmic reticulum and is involved in GPI-anchor biosynthesis. The glycosylphosphatidylinositol (GPI)-anchor is a glycolipid which contains three mannose molecules in its core backbone. The GPI-anchor is found on many blood cells and serves to anchor proteins to the cell surface. PIGM encodes a mannosyltransferase, GPI-MT-I, that transfers the first mannose to GPI on the lumenal side of the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dol-P-Man dependent GPI mannosyltransferase; DPM:GlcN-(acyl-)PI mannosyltransferase; GPI mannosyltransferase 1; GPI mannosyltransferase I; GPI-MT-I; phosphatidylinositol glycan anchor biosynthesis, class M; phosphatidylinositol glycan, class M; Phosphatidylinositol-glycan biosynthesis class M protein; PIG-M; PIG-M mannosyltransferase; PIG-M mRNA for mannosyltransferase
Gene Aliases: 4933437L05Rik; AI644422; AV302080; C920011G20Rik; GPI-MT-I; PIGM
UniProt ID: (Human) Q9H3S5, (Mouse) Q8C2R7, (Rat) Q9EQY6
Entrez Gene ID: (Human) 93183, (Mouse) 67556, (Rat) 79112
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support