Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 Extended to June 20: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MANPKLLGL ELSEAEAIGA DSARFEELLL QASKELQQAQ TTRPESTQIQ PQPGFCIKTN SSEGKVFINI CHSPSIPPPA DVTEEELLQM LEEDQAGFRI PMSLGEPHAE LDAKGQGCTA YDVAVNSDFY RRMQNSDFLR ELVITIAREG LEDKYNLQLN PEWRMMKNRP FMGSISQQNI RSEQRPRIQE LGDLYTPAPG RAESGPEKPH LNLWLEAPDL LLAEIDLPKL DGALGLSLEI GENRLVMGGP QQLYHLDAYI PLQINSHESK AAFHRKRKQL MVAMPLLLVP S (1-290 aa encoded by BC001108)
Involved in the assembly of C/D box small nucleolar ribonucleoprotein (snoRNP) particles (PubMed:17636026). Recruits the SWI/SNF complex to the core promoter of rRNA genes and enhances pre-rRNA transcription (PubMed:22368283, PubMed:24036451). Mediates interaction of TELO2 with the R2TP complex which is necessary for the stability of MTOR and SMG1 (PubMed:20864032). Positively regulates the assembly and activity of the mTORC1 complex (PubMed:24036451).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nucleolar protein 17 homolog; PIH1 domain-containing protein 1
Gene Aliases: 1110061L23Rik; 4933413A04Rik; NOP17; PIH1D1; RGD1309809
UniProt ID: (Human) Q9NWS0, (Mouse) Q9CQJ2, (Rat) Q4V7F5
Entrez Gene ID: (Human) 55011, (Mouse) 68845, (Rat) 292898
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support