Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: HDSKMRLEQD LKNQALDNRE IDKKMNSIKP DLIQLRKIRD QHLVWLNHKG VRQKRLNVWL GIKNEDAAEN YFINEEDENL PHYDEKTWFV EDINRVQAED LLYGKPDGAF
PIK3R3 binds to activated (phosphorylated) protein-tyrosine kinases through its SH2 domain and regulates their kinase activity. During insulin stimulation, it also binds to IRS-1. It is a component of a heterodimer of p110 (catalytic) and p55 (regulatory) subunits. The protein is expressed at highest levels in brain and testis. Lower levels are detected in adipose tissue, kidney, heart, lung and skeletal muscle. The protein contains 2 SH2 domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p55PIK; Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; Phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; PI3K regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; RP4-533D7.2
Gene Aliases: p55; p55-GAMMA; p55PIK; PIK3R3
UniProt ID: (Human) Q92569
Entrez Gene ID: (Human) 8503
Molecular Function:
kinase modulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support