Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MGRPLLLPLL PLLLPPAFLQ PSGSTGSGPS YLYGVTQPKH LSASMGGSVE IPFSFYYPWE LATAPDVRIS WRRGHFHGQS FYSTRPPSIH KDYVNRLFLN WTEGQKSGFL RISNLQKQDQ SVYFCRVELD TRSSGRQQWQ SIEGTKLSIT QGQQRTKATT PAREPFQNTE EPYENIRNEG QNTDPKLNPK LHLTQSTSQP PSPQEPPERD PVLCLKGLTN GQPSQD
SHP-1-mediated dephosphorylation of protein tyrosine residues is central to the regulation of several cell signaling pathways. Two types of inhibitory receptor superfamily members are immunoreceptor tyrosine-based inhibitory motif (ITIM)-bearing receptors and their non-ITIM-bearing, activating counterparts. Control of cell signaling via SHP-1 is thought to occur through a balance between PILRalpha-mediated inhibition and PILR beta-mediated activation. This particular gene encodes the ITIM-bearing member of the receptor pair, which functions in the inhibitory role.Cell signaling pathways rely on a dynamic interaction between activating and inhibiting processes. SHP-1-mediated dephosphorylation of protein tyrosine residues is central to the regulation of several cell signaling pathways. Two types of inhibitory receptor superfamily members are immunoreceptor tyrosine-based inhibitory motif (ITIM)-bearing receptors and their non-ITIM-bearing, activating counterparts. Control of cell signaling via SHP-1 is thought to occur through a balance between PILR alpha-mediated inhibition and PILRbeta-mediated activation. These paired immunoglobulin-like receptor genes are located in a tandem head-to-tail orientation on chromosome 7. This particular gene encodes the ITIM-bearing member of the receptor pair, which functions in the inhibitory role.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell surface receptor FDF03; Inhibitory receptor PILR-alpha; paired immunoglobin-like type 2 receptor alpha; Paired immunoglobulin-like type 2 receptor alpha
Gene Aliases: FDF03; PILRA
UniProt ID: (Human) Q9UKJ1
Entrez Gene ID: (Human) 29992
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support