Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLLSKINSLA HLRAAPCNDL HATKLAPGKE KEPLESQYQV GPLLGSGGFG SVYSGIRVSD NLPVAIKHVE KDRISDWGEL PNGTRVPMEV VLLKKVSSGF SGVIRLLDWF ERPDSFVLIL ERPEPVQDLF DFITERGALQ EELARSFFWQ VLEAVRHCHN CGVLHRDIKD ENILIDLNRG ELKLIDFGSG ALLKDTVYTD FDGTRVYSPP EWIRYHRYHG RSAAVWSLGI LLYDMVCGDI PFEHDEEIIR GQVFFRQRVS SECQHLIRWC LALRPSDRPT FEEIQNHPWM QDVLLPQETA EIHLHSLSPG PSK
The proto-oncogene PIM1 is upregulated in prostate cancer. PIM1, which belongs to the Serine/Threonine protein kinase family, is thought to play a role in signal transduction in blood cells. The protooncogene PIM1 encodes a protein kinase upregulated in prostate cancer. It may affect the structure or silencing of chromatin by phosphorylating HP1 gamma/CBX3. PIM1 is expressed primarily in cells of the hematopoietic and germ line lineages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Oncogene PIM1; pim-1 kinase 44 kDa isoform; pim-1 oncogene (proviral integration site 1); proto-oncogene serine/threonine-protein kinase pim-1; Serine/threonine-protein kinase pim-1
Gene Aliases: PIM; PIM1
UniProt ID: (Human) P11309
Entrez Gene ID: (Human) 5292
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support