Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SEGSNLTPA HHYPDFRFKT YAPLAFRYFR ELFGIKPDDY LYSICSEPLI ELSNPGASGS LFFVTSDDEF IIKTVQHKEA EFLQKLLPGY YMNLNQNPRT LLPKFYGLYC MQSGGINIRI VVMNNVLPRS MRMHFTYDLK GSTYKRRASR KEREKSNPTF KDLDFLQDMH EGLYFDTETY NALMKTLQRD CRVLESFKIM DYSLLLGIHF LDHSLKEKEE ETPQNVPDAK RTGMQKVLYS TAMESIQGPG KSGDGIITEN PDTMGGIPAK SHRGEKLLLF TGIIDILQSY RLMKKLEHSW (107-405 aa encoded by BC030587)
PIP5K1B lipid kinase or phosphatidylinositol-4-phosphate 5-kinase, type I, beta is a member of the phosphatidylinositol-4-phosphate 5-kinase family. PIP5K1B gene contains 17 exons and spans more than 300 kb. The seventeenth exon was found by RT-PCR, and is derived from the 3-prime untranslated region of the PRKACG gene which is located on 9q13 approximately 3 kb downstream of the STM7.I 3-prime untranslated region. The overexpression of PIP5K1B in COS-7 cells induces an increase in short actin fibers and a decrease in actin stress fibers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 68 kDa type I phosphatidylinositol 4-phosphate 5-kinase beta; Phosphatidylinositol 4-phosphate 5-kinase type I alpha; Phosphatidylinositol 4-phosphate 5-kinase type I beta; Phosphatidylinositol 4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase; phosphatidylinositol-4-phosphate 5-kinase type I alpha; phosphatidylinositol-4-phosphate 5-kinase type I beta; phosphatidylinositol-4-phosphate 5-kinase type-1 beta; phosphatidylinositol-4-phosphate 5-kinase, type 1 alpha; phosphatidylinositol-4-phosphate 5-kinase, type 1, beta; phosphatidylinositol-4-phosphate 5-kinase, type I, beta; PIP5K1-beta; PIP5KI-beta; PIP5KIalpha; PIP5KIB2 protein; PIP5KIbeta; Protein STM-7; ptdIns(4)P-5-kinase 1 beta; ptdIns(4)P-5-kinase beta; STM-7 protein; Type I phosphatidylinositol 4-phosphate 5-kinase beta; type I phosphatidylinositol-4-phosphate 5-kinase beta
Gene Aliases: MSS4; PI4P5K-I[b]; PIP5K1-beta; Pip5k1a; PIP5K1B; PIP5KIbeta; Pipk5b; STM7
UniProt ID: (Human) O14986, (Mouse) P70181, (Rat) Q5CZZ9
Entrez Gene ID: (Human) 8395, (Mouse) 18719, (Rat) 309419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support