Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: IGDVQILSSF VRDHLPDLKP EPAVIESCMY TNTPDEQFIL DRHPKYDNIV IGAGFSGHGF KLAPVVGKIL YELSMKLTPS YDLAPFRISR FPG
Metabolizes sarcosine, L-pipecolic acid and L-proline.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: L-pipecolate oxidase; L-pipecolic acid oxidase; Peroxisomal sarcosine oxidase; pipecolic acid oxidase; PSO
Gene Aliases: LPIPOX; PIPOX; PSO
UniProt ID: (Human) Q9P0Z9
Entrez Gene ID: (Human) 51268
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support