Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
FIGURE: 1 / 4
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a cytoplasmic protein that catalyzes the transfer of phosphatidylinositol and phosphatidylcholine between membranes. This transfer activity is required for COPI complex-mediated retrograde transport from the Golgi apparatus to the endoplasmic reticulum. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; PI-TP beta; PI-TP-beta; Pitp beta; PtdIns transfer protein beta; ptdInsTP beta
Gene Aliases: PI-TP-beta; PITPNB; PtdInsTP; VIB1B
UniProt ID: (Human) P48739, (Rat) P53812, (Mouse) P53811
Entrez Gene ID: (Human) 23760, (Rat) 114561, (Mouse) 56305
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support