Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: HepG2.
Predicted reactivity: Mouse (97%), Pig (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the phosphatidylinositol transfer protein family. The encoded cytoplasmic protein transfers phosphatidylinositol from one membrane compartment to another. Two transcript variants encoding distinct isoforms have been identified for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytoplasmic phosphatidylinositol transfer protein 1; M-rdgB beta; Mammalian rdgB homolog beta; RdgBbeta; retinal degeneration B beta 1; Retinal degeneration B homolog beta
Gene Aliases: M-RDGB-beta; MRDGBbeta; PITPNC1; RDGB-BETA; RDGBB; RDGBB1
UniProt ID: (Human) Q9UKF7
Entrez Gene ID: (Human) 26207
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support