Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The protein encoded by this gene is an ATP-dependent metalloprotease that degrades post-cleavage mitochondrial transit peptides. The encoded protein binds zinc and can also degrade amyloid beta A4 protein, suggesting a possible role in Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPreP; Metalloprotease 1; metalloprotease 1 (pitrilysin family); nuclear transplantation upregulated protein 1; pitrilysin metalloprotease 1; Pitrilysin metalloproteinase 1; PreP peptidasome; Presequence protease, mitochondrial
Gene Aliases: 2310012C15Rik; AA410010; KIAA1104; mKIAA1104; MP-1; MP1; Ntup1; PITRM1; PREP
UniProt ID: (Human) Q5JRX3, (Mouse) Q8K411
Entrez Gene ID: (Human) 10531, (Mouse) 69617
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support