Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
It is not recommended to aliquot this antibody.
Hiwi is a member of the Piwi /Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34- hematopoietic cells, suggesting a role for this gene in the maintenance of hematopoietic stem cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HILI; mili; Miwi like; Miwi likw; piwi homolog; piwi like homolog 1; piwi-like 1; piwi-like homolog 1; Piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1; PIWIL1L; piwil2-like protein
Gene Aliases: CT80.1; HIWI; MIWI; PIWI; PIWIL1
UniProt ID: (Human) Q96J94, (Mouse) Q9JMB7
Entrez Gene ID: (Human) 9271, (Rat) 363912, (Mouse) 57749
Molecular Function:
translation initiation factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support