Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This target displays homology in the following species: Cow: 90%; Dog: 90%; Goat: 85%; Guinea Pig: 92%; Horse: 90%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
This gene encodes a member of the cAMP-dependent protein kinase inhibitor family. The encoded protein may play a role in the protein kinase A (PKA) pathway by interacting with the catalytic subunit of PKA, and overexpression of this gene may play a role in prostate cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Jul 2012].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cAMP-dependent protein kinase (catalytic subunit binding) inhibtor 2; cAMP-dependent protein kinase inhibitor beta; cAMP-dependent protein kinase inhibitor, testis isoform; PKI-beta; protein kinase (cAMP dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, (cAMP-dependent, catalytic) inhibitor beta; protein kinase inhibitor beta, cAMP dependent, catalytic; protein kinase inhibitor beta, cAMP dependent, testis specific; protein kinase inhibitor, beta
Gene Aliases: PKINH; Prkacn2; RATPKINH
Entrez Gene ID: (Rat) 24678
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support