Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MEEGAPRQPG PSQWPPEDEK EVIRRAIQKE LKIKEGVENL RRVATDRRHL GHVQQLLRSS NRRLEQLHGE LRELHARILL PGPGPGPAEP VASGPRPWAE QLRARHLEAL RRQLHVELKV KQGAENMTHT CASGTPKERK LLAAAQQMLR DSQLKVALLR MKISSLEASG SPEPGPELLA EELQHRLHVE AAVAEGAKNV VKLLSSRRTQ DRKALAEAQA QLQESSQKLD LLRLALEQLL EQLPPAHPLR SRVTRELRAA VPGYPQPSGT PVKPTALTGT LQVRLLGCEQ LLTAVPGRSP AAALASSPSE GWLRTKAKHQ RGRGELASEV LAVLKVDNRV VGQTGWGQVA EQSWDQTFVI PLERARELEI GVHWRDWRQL CGVAFLRLED FLDNACHQLS LSLVPQGLLF AQVTFCDPVI ERRPRLQRQE RIFSKRRGQD FLRASQMNLG MAAWGRLVMN LLPPCSSPST ISPPKGCPRT PTTLREASDP ATPSNFLPKK TPLGEEMTPP PKPPRLYLPQ EPTSEETPRT KRPHMEPRTR RGPSPPASPT RKPPRLQDFR CLAVLGRGHF GKVLLVQFKG TGKYYAIKAL KKQEVLSRDE IESLYCEKRI LEAVGCTGHP FLLSLLACFQ TSSHACFVTE FVPGGDLMMQ IHEDVFPEPQ ARFYVACVVL GLQFLHEKKI IYRDLKLDNL LLDAQGFLKI ADFGLCKEGI GFGDRTSTFC GTPEFLAPEV LTQEAYTRAV DWWGLGVLLY EMLVGECPFP GDTEEEVFDC IVNMDAPYPG FLSVQGLEFI QKLLQKCPEK RLGAGEQDAE EIKVQPFFRT TNWQALLART IQPPFVPTLC GPADLRYFEG EFTGLPPALT PPAPHSLLTA RQQAAFRDFD FVSERFLEP
PKN3, a protein kinase C-related molecule, mediates malignant cell growth downstream of activated PI3K. PKN3 is required for invasive prostate cell growth as assessed by 3D cell culture assays and in an orthotopic mouse tumor model by inducible expression of short hairpin RNA (shRNA). PKN3 is regulated by PI3K at both the expression level and the catalytic activity level. Therefore, PKN3 might represent a preferred target for therapeutic intervention in cancers that lack tumor suppressor PTEN function or depend on chronic activation of PI3K.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein kinase PKN-beta; Protein-kinase C-related kinase 3; Serine/threonine-protein kinase N3
Gene Aliases: PKN3; PKNBETA; UTDP4-1
UniProt ID: (Human) Q6P5Z2
Entrez Gene ID: (Human) 29941
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support