Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: LPEKYSQNIY IQNRNIQTDN NKSIGCFGSR SRKVKEQYQD VPMPEEKSNP KGVEWLWHSI VIRMYLSLIA KSVRNYTQEA SLGALQNLTA GSGPMPTSVA QTVVQKESGL QHTRKMLHVG DPSVKKTAIS LLRNLSRNLS LQNEIAKETL PDLVSIIPDT VPSTDLLIET TASACYTLNN IIQNSYQNAR DLLNTGGIQK IMAISAGDAY ASNKASKAAS VLLYSLWAHT ELHHAYKKAQ FKKTDFVNSR TAKAYHSLKD
Plakophilins 1, 2, 3 and 4 (PKP1-4) influence development and participate in linking cadherins to cytoskeletal intermediate filaments. Plakophilins 1-4 contain arm-repeat (armadillo) domains and localize to nuclei and cell desmosomes (cell-cell junctions found in suprabasal layers of stratifying epithelia that undergo mechanical stress). Plakophilin 1 mediates increases in desmosomal protein content, desmosome assembly and regulation of cell migration. Plakophilin 2 is important for desmosome assembly and is an essential morphogenic factor and architectural component of the heart. Plakophilin 4 is a component of desmosomal adhesion plaques that regulates junctional plaque organization and cadherin function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Plakophilin-2
Gene Aliases: 1200008D14Rik; 1200012P04Rik; AA516617; ARVD9; PKP2; Pkp2l
UniProt ID: (Human) B8QFA1, (Mouse) Q9CQ73, (Rat) F1M7L9
Entrez Gene ID: (Human) 5318, (Mouse) 67451, (Rat) 287925
Molecular Function:
intermediate filament binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support