Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 6
Antibody detects endogenous levels of total PLA2G2A.
The protein encoded by this gene is a member of the phospholipase A2 family (PLA2). PLA2s constitute a diverse family of enzymes with respect to sequence, function, localization, and divalent cation requirements. This gene product belongs to group II, which contains secreted form of PLA2, an extracellular enzyme that has a low molecular mass and requires calcium ions for catalysis. It catalyzes the hydrolysis of the sn-2 fatty acid acyl ester bond of phosphoglycerides, releasing free fatty acids and lysophospholipids, and thought to participate in the regulation of the phospholipid metabolism in biomembranes. Several alternatively spliced transcript variants with different 5' UTRs have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 3.1.1.4; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; non-pancreatic secreted type II phospholipase A2; Non-pancreatic secretory phospholipase A2; NPS-PLA2; Phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2, group IIA (platelets, synovial fluid); Phospholipase A2, membrane associated; PLA2BPLA2; PLA2LPLA2S; platelet phospholipase A2; secretory group II phospholipase A2
Gene Aliases: EF; MOM1; PLA2; PLA2B; PLA2G2A; PLA2L; PLA2S; PLAS1; RASF-A; sPLA2; sPla2-IIA
UniProt ID: (Human) P14555, (Rat) P14423, (Mouse) P31482
Entrez Gene ID: (Human) 5320, (Rat) 29692, (Mouse) 18780
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support