Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: 293T, A431, Raji.
Predicted reactivity: Human (99%), Bovine (86%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calcium-dependent phospholipase A2 that selectively hydrolyzes glycerophospholipids in the sn-2 position. Has higher enzyme activity for phosphatidylethanolamine than phosphatidylcholine.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3.1.1.4; cPLA2-zeta; Cytosolic phospholipase A2 zeta; Phospholipase A2 group IVF; phospholipase A2, group IVF
Gene Aliases: PLA2G4F; PLA2G4FZ
UniProt ID: (Human) Q68DD2
Entrez Gene ID: (Human) 255189
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support