Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 8
Immunogen sequence: EEKTWHEALR SCQADNSALI DITSLAEVEF LVTLLGDENA SETWIGLSSN KIPVSFEWSN DSSVIFTNWH TLEPHIFPNR SQLCVSAEQS EGHWKVKNCE ERLFYICKKA GHVLSDAESG CQEGWERHGG FCYKID
Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 76%.
Binds to an epitope located within the peptide sequence KIPVSFEWSN as determined by overlapping synthetic peptides.
Receptor for secretory phospholipase A2 (sPLA2). Acts as a receptor for phosholipase sPLA2-IB/PLA2G1B but not sPLA2-IIA/PLA2G2A. Also able to bind to snake PA2-like toxins. Although its precise function remains unclear, binding of sPLA2 to its receptor participates in both positive and negative regulation of sPLA2 functions as well as clearance of sPLA2. Binding of sPLA2-IB/PLA2G1B induces various effects depending on the cell type, such as activation of the mitogen-activated protein kinase (MAPK) cascade to induce cell proliferation, the production of lipid mediators, selective release of arachidonic acid in bone marrow-derived mast cells. In neutrophils, binding of sPLA2-IB/PLA2G1B can activate p38 MAPK to stimulate elastase release and cell adhesion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 180 kDa secretory phospholipase A2 receptor; C-type lectin domain family 13 member C; M-type receptor; phospholipase A2 receptor 1, 180kD; phospholipase A2 receptor 1, 180kDa; PLA2-R; Secretory phospholipase A2 receptor
Gene Aliases: CLEC13C; PLA2-R; PLA2G1R; PLA2IR; PLA2R; PLA2R1
UniProt ID: (Human) Q13018
Entrez Gene ID: (Human) 22925
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support