Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MLLNRPLDG VTNSLPSPEQ LKGKILLKGK KLGGLLPPGG EGGPEATVVS DEDEAAEMED EAVRSRVQHK PKEDKLRLAQ ELSDMVIYCK SVHFGGFSSP GTPGQAFYEM ASFSENRALR LLQESGNGFV RHNVGHLSRI YPAGWRTDSS NYSPVEMWNG GCQIVALNFQ TPGPEMDVYQ GRFQDNGACG YVLKPAFLRD PNGTFNPRAL AQGPWWARKR LNIRVISGQQ LPKVNKNKNS IVDPKVTVEI HGVSRDVASR QTAVITNNGF NPWWDTEFAF EVVVPDLALI RFLVEDYDAS SKNDFIGQST IPLNSLKQGY RHVHLMSKNG DQHPSATLFV KISLQD (412-756 aa encoded by BC050382)
Phosphoinositide-specific phospholipase C (PLC) acts as a signal transducer that generates 2 second messengers, diacylglycerol and inositol 1, 4, 5-trisphosphate, by hydrolyzing inositol phospholipids. PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1; Phosphoinositide phospholipase C-delta-1; phospholipase C, delta 1; Phospholipase C-delta-1; Phospholipase C-III; PLC-delta-1; PLC-III
Gene Aliases: NDNC3; PLC-III; PLCD1
UniProt ID: (Human) P51178
Entrez Gene ID: (Human) 5333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support