Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Recommended positive controls: A549, HeLa, HepG2, HCT116.
Predicted reactivity: Mouse (86%), Rat (86%), Pig (92%), Bovine (91%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosphoinositide-specific phospholipase C (PLC) acts as a signal transducer that generates 2 second messengers, diacylglycerol and inositol 1, 4, 5-trisphosphate, by hydrolyzing inositol phospholipids. PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 ).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1; Phosphoinositide phospholipase C-delta-1; phospholipase C, delta 1; Phospholipase C-delta-1; Phospholipase C-III; PLC-delta-1; PLC-III
Gene Aliases: NDNC3; PLC-III; PLCD1
UniProt ID: (Human) P51178
Entrez Gene ID: (Human) 5333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support