Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Inactive phospholipase C-like protein 2; phospholipase C; phospholipase C, epsilon 2; Phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLC-epsilon-2; PLC-L(2); PLC-L2
Gene Aliases: KIAA1092; mKIAA1092; PLC-L2; PLCE2; PLCL2; PRIP-2
UniProt ID: (Human) Q9UPR0, (Mouse) Q8K394
Entrez Gene ID: (Pig) 100625981, (Human) 23228, (Mouse) 224860, (Rat) 301173
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support