Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LRDPICDDFF QLWQDMAESN ANIYEQIFRC LPSNATRSLR TLREYVAVEP LATVSPPLAR SELTQVQGHL VHFPLKFLED ESLLPPLGSK EGMIPLEVWT
PLD2 is a peripheral membrane protein belonging to phospholipase D family with a PH domain, two PLD phosphodiesterase domains and a PX (phox homology) domain that plays a regulatory rather than catalytic role. PLD2 catalyzes the hydrolysis of phosphatidylcholine (PC) to produce phosphatidic acid and choline. PLD2 may function in regulated secretion, signal-induced cytoskeletal regulation and/or endocytosis, transcriptional regulation, and cell cycle control. It is usually stimulated by phosphatidylinositol 4,5-bisphosphate (PIP2) and activated by the ADP-ribosylation factor-1 (ARF-1). PLD2 is activated by agonist stimulation of both tyrosine kinase and G protein-coupled receptors and is also known to interact with EGFR and PIP5K1A. PLD2, localized to plasma membrane caveolae, is ubiquitously expressed in most tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Choline phosphatase 2; hPLD2; Phosphatidylcholine-hydrolyzing phospholipase D2; phospholipase D gene 2; Phospholipase D2; PLD 2; PLD1C; rPLD2
Gene Aliases: PLD2; Pldc
UniProt ID: (Human) O14939, (Rat) P70498
Entrez Gene ID: (Human) 5338, (Rat) 25097
Molecular Function: phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support