Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPPRRPWDRE AGTLQVLGAL AVLWLGSVAL ICLLWQVPRP PTWGQVQPKD VPRSWEHGSS PAWEPLEAEA RQQRDSCQLV LVESIPQDLP SAAGSPSAQP LGQAWLQLLD TAQESVHVAS YYWSLTGPDI GVNDSSSQLG EALLQKLQQL LGRNISLAVA TSSPTLARTS TDLQVLAARG AHVRQVPMGR LTRGVLHSKF WVVDGRHIYM GSANMDWRSL TQVKELGAVI YNCSHLAQDL EKTFQTYWVL GVPKAVLPKT WPQNFSSHFN RFQPFHGLFD GVPTTAYFSA SPPALCPQGR TRDLEALLAV MGSAQEFIYA SVMEYFPTTR FSHPPRYWPV LDNALRAAAF GKGVRVRLLV GCGLNTDPTM FPYLRSLQAL SNPAANVSVD VKVFIVPVGN HSNIPFSRVN HSKFMVTEKA AYIGTSNWSE DYFSSTAGVG LVVTQSPGAQ PAGATVQEQL RQLFERDWSS RYAVGLDGQA PGQDCVWQG
PLD4 (Phospholipase D Family Member 4) is a Protein Coding gene. Among its related pathways are Innate Immune System and Inositol phosphate metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-3' exonuclease PLD4; Choline phosphatase 4; Phosphatidylcholine-hydrolyzing phospholipase D4; Phospholipase D family member 4; phospholipase D family, member 4; Phospholipase D4; PLD 4
Gene Aliases: C14orf175; PLD4; UNQ2488/PRO5775
UniProt ID: (Human) Q96BZ4
Entrez Gene ID: (Human) 122618
Molecular Function:
phospholipase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support