Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FVVCAECSR QRFLLPRLSP KPVRVCSLCY RELAAQQRQE EAEEQGAGSP GQPAHLARPI CGASSGDDDD SDEDKEGSRD GDWPSSVEFY ASGVAWSAFH S (180-297 aa encoded by BC002744)
APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AI037048; Apoptosis-inducing protein; apoptosis-inducing protein D; Lysosome-associated apoptosis-inducing protein containing PH and FYVE domains; PH and FYVE domain-containing protein 1; PH domain-containing family F member 1; phafin 1; Phafin-1; pleckstrin homology domain containing, family F (with FYVE domain) member 1; Pleckstrin homology domain-containing family F member 1; Zinc finger FYVE domain-containing protein 15
Gene Aliases: 1810013P09Rik; APPD; LAPF; PHAFIN1; PLEKHF1; ZFYVE15
UniProt ID: (Human) Q96S99, (Mouse) Q3TB82
Entrez Gene ID: (Human) 79156, (Mouse) 72287
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support