Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MMKKNNSAKR GPQDGNQQPA PPEKVGWVRK FCGKGIFREI WKNRYVVLKG DQLYISEKEV KDEKNIQEVF DLSDYEKCEE LRKSKSRSKK NHSKFTLAHS KQPGNTAPNL IFLAVSPEEK ESWINALNSA ITRAKNRILD EVTVEEDSYL AHPTRDRAKI QHSRRPPTRG HLMAVASTST SDGMLTLDLI QEEDPSPEEP TSCAESFRVD LDKSVAQLAG SRRRADSDRI QPSADRASSL SRPWEKTDKG ATYTPQAPKK LTPTEKGRCA SLEEILSQRD AASARTLQLR AEEPPTPALP NPGQLSRIQD LVARKLEETQ ELLAEVQGLG DGKRKAKDPP RSPPDSESEQ LLLETERLLG EASSNWSQAK RVLQEVRELR DLYRQMDLQT PDSHLRQTTP HSQYRKSLM
CKIP1 was identified through a yeast-two hybrid screening as a protein that would interact with only one of the two catalytic subunits of the casein kinase 2 complex. CKIP1 is a pleckstrin homology domain-containing protein localized within the nucleus and at the plasma membrane that interacts with CK2alpha but not CK2alpha', and is thought to play a role in targeting CK2alpha to a particular cellular location. CKIP1 has been implicated in muscle differentiation and the regulation of cell morphology and actin cytoskeleton. CKIP1 can also interact with other proteins such as ATM, an upstream kinase of p53, and recruit the nuclear ATM to the plasma membrane, suggesting CKIP1 may regulate ATM function through re-localizing ATM. CKIP1 can also form a complex with the kinase AKT, leading to a down-regulation of PI3K/AKT signaling and suppression of tumor growth in vivo.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C-Jun-binding protein; Casein kinase 2-interacting protein 1; CK2 interacting protein 1; HQ0024c protein; CK2-interacting protein 1; HQ0024c protein; JBP; Osteoclast maturation-associated gene 120 protein; PH domain-containing family O member 1; pleckstrin homology domain containing, family O member 1; Pleckstrin homology domain-containing family O member 1; RP11-458I7.3
Gene Aliases: CKIP-1; CKIP1; HQ0024c; JBP; OC120; PLEKHO1
UniProt ID: (Human) Q53GL0
Entrez Gene ID: (Human) 51177
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support