Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAVNKGLTLL DGDLPEQENV LQRVLQLPVV SGTCECFQKT YTSTKEAHPL VASVCNAYEK GVQSASSLAA WSMEPVVRRL STQFTAANEL ACRGLDHLEE KIPALQYPPE KIASELKDTI STRLRSARNS ISVPIASTSD KVLGAALAGC ELAWGVARDT AEFAANTRAG RLASGGADLA LGSIEKVVEY LLPADKEESA PAPGHQQAQE SPKAKPSLLS RVGALTNTLS RYTVQTMARA LEQGHTVAMW IPGVVPLSSL AQWGASVAMQ AVSRRRSEVR VPWLHSLAAA QEEDHEDQTD TEGEDTEEEE ELETEENKFS EVAALPGPRG LLGGVAHTLQ KTLQTTISAV TWAPAAVLGM AGRVLHLTPA PAVSSTKGRA MSLSDALKGV TDNVVDTVVH YVPLPRLSLM EPESEFRDID NPPAEVERRE AERRASGAPS AGPEPAPRLA QPRRSLRSAQ SPGAPPGPGL EDEVATPAAP RPGFPAVPRE KPKRRVSDSF FRPSVMEPIL GRTHYSQLRK KS
Adipose tissue is an energy reserve in animals and is strictly regulated in nondomestic species. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), resistin, and perilipin. Perilipin is an intracellular neutral lipid droplet protein that is hormonally regulated. This protein is localized exclusively to the surface of lipid droplets. In response to lypotic stimuli, perilipin is phosphorylated by protein kinase A. Once activated, perilipin has inhibitory affects upon hormone-sensitive lipase (HSL), a protein that mediates the hydrolysis of triacylglycerol, the major form of stored energy in the body. Perilipin expression is limited to adipocytes and steroidogenic cells. There are currently two known isoforms, Perilipin A and B. Both of these proteins are encoded by a single-copy gene and are the result of differential splicing events.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lipid droplet-associated protein; perilipin; Perilipin-1
Gene Aliases: FPLD4; PERI; PLIN; PLIN1
UniProt ID: (Human) O60240
Entrez Gene ID: (Human) 5346
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support