Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bone protein I (BP-I); LH3; lysine hydroxylase 3; lysyl hydroxlase 3; lysyl hydroxylase 2; lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3; procollagen-lysine,2-oxoglutarate 5-dioxygenase 3
Gene Aliases: AI414586; LH3; PLOD3
UniProt ID: (Human) O60568, (Mouse) Q9R0E1, (Rat) Q5U367
Entrez Gene ID: (Human) 8985, (Mouse) 26433, (Rat) 288583
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support