Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to PLS1. This antibody is specific to PLS1. It has no cross reaction to PLS3.
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. The protein encoded by this gene is a third distinct plastin isoform, which is specifically expressed at high levels in the small intestine. Alternatively spliced transcript variants varying in the 5' UTR, but encoding the same protein, have been found for this gene. A pseudogene of this gene is found on chromosome 11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fimbrin; I plastin; I-fimbrin; I-plastin; intestine specific plastin; Intestine-specific plastin; plastin 1 (I isoform); plastin 1, I; Plastin-1
Gene Aliases: AI427122; PLS1
UniProt ID: (Human) Q14651, (Mouse) Q3V0K9
Entrez Gene ID: (Human) 5357, (Rat) 315926, (Mouse) 102502
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support