Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MDEMATTQIS KDELDELKEA FAKVDLNSNG FICDYELHEL FKEANMPLPG YKVREIIQKL MLDGDRNKDG KISFDEFVYI FQEVKSSDIA KTFRKAINRK EGICALGGTS ELSSEGTQHS YSEEEKYAFV NWINKALEND PDCRHVIPMN PNTDDLFKAV GDGIVLCKMI NLSVPDTIDE RAINKKKLTP FIIQENLNLA LNSASAIGCH VVNIGAEDLR AGKPHLVLGL LWQIIKIGLF ADIELSRNEA LAALLRDGET LEELMKLSPE ELLLRWANFH LENSGWQKIN NFSADIKDSK AYFHLLNQIA PKGQKEGEPR IDINMSGFNE TDDLKRAESM LQQADKLGCR QFVTPADVVS GNPKLNLAFV ANLFNKYPAL TKPENQDIDW TLLEGETREE RTFRNWMNSL GVNPHVNHLY ADLQDALVIL QLYERIKVPV DWSKVNKPPY PKLGANMKKL ENCNYAVELG KHPAKFSLVG IGGQDLNDGN QTLTLALVWQ LMRRYTLNVL EDLGDGQKAN DDIIVNWVNR TLSEAGKSTS IQSFKDKTIS SSLAVVDLID AIQPGCINYD LVKSGNLTED DKHNNAKYAV SMARRIGARV YALPEDLVEV KPKMVMTVFA CLMGRGMKRV
Phospholipid scramblases (PLS) translocate phospholipids bidirectionally between inner and outer leaflets of the plasma membrane and are implicated in apoptosis (1,2). Members of this family, PLS1 and PLS3 are both substrates of Protein kinase C (PKC) delta and are phosphorylated by PKC-delta during apoptosis. PKC-delta translocates to mitochondria during apoptosis and phosphorylates PLS3. Overexpression of PLS3 in the HEK293 cells enhanced apoptosis induced by UV-irradiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Plastin-3; T fimbrin; T plastin; T-plastin
Gene Aliases: BMND18; PLS3; T-plastin
UniProt ID: (Human) P13797
Entrez Gene ID: (Human) 5358
Molecular Function: non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support