Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: 293T, Mouse brain, Rat brain.
Immunogen sequence: LDTVPVRSSV DELVGIDYSL MKDPVASTSN LDMDFRGAFF PLTERNWSLP NRAVEPQLQE EERMVYVAFS EFFFDSAMES YFRAGALQLL LVGDKVPHDL D
PLTP is one of at least two lipid transfer proteins found in human plasma. The protein transfers phospholipids from triglyceride-rich lipoproteins to high density lipoprotein (HDL). In addition to regulating the size of HDL particles, this protein may be involved in cholesterol metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BPI fold containing family E; Lipid transfer protein II; Phospholipid transfer protein
Gene Aliases: BPIFE; HDLCQ9; OD107; PLTP
UniProt ID: (Human) P55058, (Mouse) P55065
Entrez Gene ID: (Human) 5360, (Mouse) 18830, (Rat) 296371
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support