Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 1
In direct ELISAs, no cross-reactivity with recombinant human Plexin A1, recombinant mouse (rm) Plexin A1 or rmPlexin A2 is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
PLXNA4 is a protein coding gene. Coreceptor for SEMA3A. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000207245; Plexin-A4
Gene Aliases: FAYV2820; KIAA1550; PLEXA4; PLXNA4; PLXNA4A; PLXNA4B; PRO34003; UNQ2820/PRO34003
UniProt ID: (Human) Q9HCM2
Entrez Gene ID: (Human) 91584
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support