Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PMM1, also named as PMMH22, belongs to the eukaryotic PMM family. It catalyzes the second step in the conversion of fructose-6P to GDP-mannose is and is involved in the synthesis of the GDP-mannose and dolichol-phosphate-mannose required for a number of critical mannosyl transfer reactions. In addition, PMM1 may be responsible for the degradation of glucose-1, 6-bisphosphate in ischemic brain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brain glucose-1,6-bisphosphatase; Phosphomannomutase 1; PMM 1; PMMH-22; Secp53 homolog
Gene Aliases: C77612; PMM1; PMMH22; Sec53
UniProt ID: (Human) A8K003, (Mouse) O35621
Entrez Gene ID: (Human) 5372, (Mouse) 29858, (Rat) 300089
Molecular Function: mutase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support