Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: IQNSTLGEAE IERERGVILR EMQEVETNLQ EVVFDYLHAT AYQNTALGRT ILGPTENIKS ISRKDLVDYI TTHYKGPRIV LAAAGGVSHD ELLDLAKFHF GDSLCTHKG
PMPCB is a member of the peptidase M16 family and encodes a protein with a zinc-binding motif. This protein is located in the mitochondrial matrix and catalyzes the cleavage of the leader peptides of precursor proteins newly imported into the mitochondria, though it only functions as part of a heterodimeric complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-MPP; mitochondrial processing peptidase beta subunit; Mitochondrial-processing peptidase subunit beta; P-52; peptidase (mitochondrial processing) beta
Gene Aliases: Beta-MPP; MPP11; MPPB; MPPP52; P-52; PMPCB
UniProt ID: (Human) O75439
Entrez Gene ID: (Human) 9512
Molecular Function:
metalloprotease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support