Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 3
Recommended positive controls: 293T, A431, HeLa, HepG2.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein belonging to the DNA mismatch repair mutL/hexB family. This protein is thought to be involved in the repair of DNA mismatches, and it can form heterodimers with MLH1, a known DNA mismatch repair protein. Mutations in this gene cause hereditary nonpolyposis colorectal cancer type 3 (HNPCC3) either alone or in combination with mutations in other genes involved in the HNPCC phenotype, which is also known as Lynch syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA mismatch repair protein PMS1; human homolog of yeast mutL; mismatch repair gene PMSL1; PMS1 postmeiotic segregation increased 1; PMS1 protein homolog 1; rhabdomyosarcoma antigen MU-RMS-40.10B; rhabdomyosarcoma antigen MU-RMS-40.10E
Gene Aliases: HNPCC3; hPMS1; MLH2; PMS1; PMSL1
UniProt ID: (Human) P54277
Entrez Gene ID: (Human) 5378
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support