Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SHCSSEYAA SSPGDRGSQE HVDSQEKAPE TDDSFSDVDC HSNQEDTGCK FRVLPQPTNL ATPNTKRFKK EEILSSSDIC QKLVNTQDMS ASQVDVAVKI NKKVVPLDFS ISSLAKRIKQ LHHEAQQSEG EQNYRKFRAK ICPGENQAAE DELRKEISKT MFAEMEIIGQ FNLGFIITKL NEDIFIVDQH ATDEKYNFEM LQQHTVLQGQ RLIAPQTLNL TAVNEAVLIE NLEIFRKNGF DFVIDENAPV TERAKLISLP TSKNWTFGPQ DVDELIFMLS DSPGVMCRPS RVKQMFASRA CRKSVMIGTA LNTSEMKKLI THMGEMDHPW NCPHGRPTMR HIANLGVISQ N (513-862 aa encoded by BC093921)
PMS1 protein homologue 2 (PMS2) is a DNA mismatch repair protein. Defects in PMS2 are a cause of hereditary non-polyposis cancer (HNPCC). Human PMS2 is an amino acid protein of 862 amino acids.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA mismatch repair protein PMS2; H_DJ0042M02.9; Mismatch repair endonuclease PMS2; PMS1 homolog 2, mismatch repair protein; PMS1 protein homolog 2; PMS2 postmeiotic segregation increased 2; postmeiotic segregation increased 2 nirs variant 6
Gene Aliases: AW555130; HNPCC4; MLH4; PMS2; PMS2CL; PMSL2
UniProt ID: (Human) P54278, (Mouse) P54279
Entrez Gene ID: (Human) 5395, (Rat) 288479, (Mouse) 18861
Molecular Function: DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support