Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAPLGGAPR LVLLFSGKRK SGKDFVTEAL QSRLGADVCA VLRLSGPLKE QYAQEHGLNF QRLLDTSTYK EAFRKDMIRW GEEKRQADPG FFCRKIVEGI SQPIWLVSDT RRVSDIQWFR EAYGAVTQTV RVVALEQSRQ QRGWVFTPGV DDAESECGLD NFGDFDWVIE NHGVEQRLEE QLENLIEFIR SRL (1-192 aa encoded by BC007694)
PMVK is a peroxisomal enzyme that catalyzes the conversion of mevalonate 5-phosphate into mevalonate 5-diphosphate as the fifth reaction of the cholesterol biosynthetic pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPMK; Phosphomevalonate kinase; PMKase; testis tissue sperm-binding protein Li 95mP
Gene Aliases: HUMPMKI; PMK; PMKA; PMKASE; PMKI; PMVK; POROK1
UniProt ID: (Human) Q15126
Entrez Gene ID: (Human) 10654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support