Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Human patatin-like phospholipases, such as PNPLA1, have been implicated in regulation of adipocyte differentiation and have been induced by metabolic stimuli (Wilson et al., 2006 [PubMed 16799181]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: patatin-like phospholipase domain-containing protein 1
Entrez Gene ID: (Rat) 361812
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support