Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVQMMRQFLY RVLPEDSYKV TTGKLHVSLT RLTDGENVVV SEFTSKEELI EALYCSCFVP VYCGLIPPTY RGVWAFLTLP PQRYIDGGFT GMQPCAFWTD AITISTFSGQ QDICPRDCPA IFHDFRMFNC SFQFSLENIA RMTHALFPPD LVILHDYYYR GYEDAVLYLR RLNAVYLNSS SKRVIFPRVE VYCQIELALG NECPERSQPS LRARQASLEG ATQPHKEWVP KGDGRGSHGP PVSQPVQTLE FTCESPVSAP VSPLEQPPAQ PLASSTPLSL SGMPPVSFPA VHKPPSSTPG SSLPTPPPGL SPLSPQQQVQ PSGSPARSLH SQAPTSHRPS LGPSTVGAPQ TLPRSSLSAF PAQPPVEELG QEQPQAVALL VSSKPKSAVP LVHVKETVSK PYVMESPAED SNWVNKVFKK NKQKTSGTRK GFPRHPGSKK PSSKVQ
Human patatin-like phospholipases, such as PNPLA1, have been implicated in regulation of adipocyte differentiation and have been induced by metabolic stimuli (Wilson et al., 2006 [PubMed 16799181]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Omega-hydroxyceramide transacylase; patatin-like phospholipase domain containing 1; Patatin-like phospholipase domain-containing protein 1
Gene Aliases: ARCI10; dJ50J22.1; PNPLA1
UniProt ID: (Human) A3RMU3
Entrez Gene ID: (Human) 285848
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support