Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTCWLRGVT ATFGRPAEWP GYLSHLCGRS AAMDLGPMRK SYRGDREAFE ETHLTSLDPV KQFAAWFEEA VQCPDIGEAN AMCLATCTRD GKPSARMLLL KGFGKDGFRF FTNFESRKGK ELDSNPFASL VFYWEPLNRQ VRVEGPVKKL PEEEAECYFH SRPKSSQIGA VVSHQSSVIP DREYLRKKNE ELEQLYQDQE VPKPKSWGGY VLYPQVMEFW QGQTNRLHDR IVFRRGLPTG DSPLGPMTHR GEEDWLYERL AP (1-261 aa encoded by BC006525)
The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5'-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 302; FLJ10535; pyridoxal 5'-phosphate synthase; Pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; Pyridoxine-5'-phosphate oxidase
Gene Aliases: AI415282; HEL-S-302; PDXPO; PNPO
UniProt ID: (Human) Q9NVS9, (Rat) O88794, (Mouse) Q91XF0
Entrez Gene ID: (Human) 55163, (Rat) 64533, (Mouse) 103711
Molecular Function: oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support