Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Immunogen sequence: MTCWLRGVTA TFGRPAEWPG YLSHLCGRSA AMDLGPMRKS YRGDREAFEE THLTSLDPVK QFAAWFEEAV QCPDIGEANA MCLATCTRDG KPSARMLLLK GFGKDGFRFF TNFESRKGKE LDSNPFASLV FYWEPLNRQV RVEGPVKKLP EEEAECYFHS RPKSSQIGAV VSHQSSVIPD REYLRKKNEE LEQLYQDQEV PKPKSWGGYV LYPQVMEFWQ GQTNRLHDRI VFRRGLPTGD SPLGPMTHRG EEDWLYERLA P
The enzyme encoded by this gene catalyzes the terminal, rate-limiting step in the synthesis of pyridoxal 5'-phosphate, also known as vitamin B6. Vitamin B6 is a required co-factor for enzymes involved in both homocysteine metabolism and synthesis of neurotransmitters such as catecholamine.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis secretory protein Li 302; FLJ10535; pyridoxal 5'-phosphate synthase; Pyridoxamine-phosphate oxidase; pyridoxine 5'-phosphate oxidase; pyridoxine 5-phosphate oxidase; Pyridoxine-5'-phosphate oxidase
Gene Aliases: AI415282; HEL-S-302; PDXPO; PNPO
UniProt ID: (Human) Q9NVS9, (Rat) O88794, (Mouse) Q91XF0
Entrez Gene ID: (Human) 55163, (Rat) 64533, (Mouse) 103711
Molecular Function: oxidase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support