Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody detected the endogenous PNPT1 around 80 kDa in mouse brain.
Immunogen sequence: YEFPPYATN EIGKVTGLNR RELGHGALAE KALYPVIPRD FPFTIRVTSE VLESNGSSSM ASACGGSLAL MDSGVPISSA VAGVAIGLVT KTDPEKGEIE DYRLLTDILG IEDYNGDMDF KIAGTNKGIT ALQADIKLPG IPIKIVMEAI QQASVAKKEI LQIMNKTISK PRASRKENGP VVETVQVPLS KRAKFVGPGG YNLKKLQAET GVTISQVDEE TFSVFAPTPS AMHEARDFIT EICKDDQEQQ LEFGAVYTAT ITEIRDTGVM VKLYPNMTAV LLHNTQLDQR KIKHPTALGL EVGQEIQVKY FGRDPADGRM RLSRKVLQSP ATTVVRTLND RSSIVMGEPI SQSSSNSQ (427-783 aa encoded by BC053660)
PNPT1 (polyribonucleotide nucleotidyltransferase 1, mitochondirla) is a RNA-binding protein implicated in numerous RNA metabolic processes. It catalyzes the phosphorolysis of single-stranded polyribonucleotides processively in the 3'-to-5' direction. PNPT1 is a component of the mitochondrial degradosome (mtEXO) complex that degrades 3' overhang double-stranded RNA with a 3'-to-5' directionality in an ATP-dependent manner. It is required for correct processing and polyadenylation of mitochondrial mRNAs. It plays a role as a cytoplasmic RNA import factor, in mitochondrial morphogenesis and respiration, regulation of expression of the electron transport chain, regulation of the stability of specific mature miRNAs in melanoma cells, and RNA cell surveilance. Mutations of the gene can result in combined oxidative phosphorylation deficiency 13 (COXPD13) and deafness, autosomal recessive, 70 (DFNB70).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3'-5' RNA exonuclease OLD35; DKFZp762K1914; old 35; PNPase 1; PNPase old-35; Polynucleotide phosphorylase 1; Polynucleotide phosphorylase-like protein; Polyribonucleotide nucleotidyltransferase 1, mitochondrial
Gene Aliases: 1200003F12Rik; COXPD13; DFNB70; old-35; OLD35; PNPASE; PNPT1; Pnptl1
UniProt ID: (Human) Q8TCS8, (Mouse) Q8K1R3
Entrez Gene ID: (Human) 87178, (Rat) 360992, (Mouse) 71701
Molecular Function:
nucleotidyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support