Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SWDPAGYLLY CPCMGRFGNQ ADHFLGSLAF AKLLNRTLAV PPWIEYQHHK PPFTNLHVSY QKYFKLEPLQ AYHRVISLED FMEKLAPTHW PPEKRVAYCF EVAAQRSPDK KTCPMKEGNP FGPFWDQFHV SFNKSELFTG ISFSASYREQ WSQRRENHSC VTLLFPR; Positive Samples: HepG2, HL-60, Mouse heart, Mouse liver; Cellular Location: Endoplasmic reticulum
POFUT1, an endoplasmic reticulum-residing member of the glycosyltransferase O-Fuc family, adds O-fucose through an O-glycosidic linkage to conserved serine or threonines in epidermal growth factor-like repeats of several cell surface and secreted proteins. Unlike its homolog POFUT2, POFUT1 can also catalyze the transfer of fucose to thrombospondin type 1 repeats. Many of the substrates of POFUT1 are involved in ligand-induced receptor signaling. One such protein is Notch; mouse ES cells lacking POFUT have normal levels of Notch receptors at the cell surface, but these receptors do not bind Notch ligands or exhibit Notch signaling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDP-fucose protein O-fucosyltransferase 1; o-fucosyltransferase protein; O-FucT-1; Peptide-O-fucosyltransferase 1; pofut1 {ECO:0000312|RGD:1303001}
Gene Aliases: DDD2; FUT12; KIAA0180; mKIAA0180; O-Fuc-T; O-FucT-1; O-FUT; OFUCT1; POFUT1
UniProt ID: (Human) Q9H488, (Mouse) Q91ZW2, (Rat) Q6EV70
Entrez Gene ID: (Human) 23509, (Mouse) 140484, (Rat) 311551
Molecular Function: protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support